Merck
A TNIP1-driven systemic autoimmune disorder with elevated IgG4.
Nature immunology (2024-07-27)
Arti Medhavy, Vicki Athanasopoulos, Katharine Bassett, Yuke He, Maurice Stanley, Daniel Enosi Tuipulotu, Jean Cappello, Grant J Brown, Paula Gonzalez-Figueroa, Cynthia Turnbull, Somasundhari Shanmuganandam, Padmaja Tummala, Gemma Hart, Tom Lea-Henry, Hao Wang, Sonia Nambadan, Qian Shen, Jonathan A Roco, Gaetan Burgio, Phil Wu, Eun Cho, T Daniel Andrews, Matt A Field, Xiaoqian Wu, Huihua Ding, Qiang Guo, Nan Shen, Si Ming Man, Simon H Jiang, Matthew C Cook, Carola G Vinuesa
ABSTRACT
Whole-exome sequencing of two unrelated kindreds with systemic autoimmune disease featuring antinuclear antibodies with IgG4 elevation uncovered an identical ultrarare heterozygous TNIP1Q333P variant segregating with disease. Mice with the orthologous Q346P variant developed antinuclear autoantibodies, salivary gland inflammation, elevated IgG2c, spontaneous germinal centers and expansion of age-associated B cells, plasma cells and follicular and extrafollicular helper T cells. B cell phenotypes were cell-autonomous and rescued by ablation of Toll-like receptor 7 (TLR7) or MyD88. The variant increased interferon-β without altering nuclear factor kappa-light-chain-enhancer of activated B cells signaling, and impaired MyD88 and IRAK1 recruitment to autophagosomes. Additionally, the Q333P variant impaired TNIP1 localization to damaged mitochondria and mitophagosome formation. Damaged mitochondria were abundant in the salivary epithelial cells of Tnip1Q346P mice. These findings suggest that TNIP1-mediated autoimmunity may be a consequence of increased TLR7 signaling due to impaired recruitment of downstream signaling molecules and damaged mitochondria to autophagosomes and may thus respond to TLR7-targeted therapeutics.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-HA antibody, Mouse monoclonal, clone HA-7, purified from hybridoma cell culture
Sigma-Aldrich
Anti-TNIP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-EEA1 antibody, Mouse monoclonal, clone EEA1-N19, purified from hybridoma cell culture
Sigma-Aldrich
Anti-HA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution