Merck
  • Human cortical interneurons optimized for grafting specifically integrate, abort seizures, and display prolonged efficacy without over-inhibition.
Human cortical interneurons optimized for grafting specifically integrate, abort seizures, and display prolonged efficacy without over-inhibition.
Neuron (2023-01-11)
Qian Zhu, Akanksha Mishra, Joy S Park, Dongxin Liu, Derek T Le, Sasha Z Gonzalez, Morgan Anderson-Crannage, James M Park, Gun-Hoo Park, Laura Tarbay, Kamron Daneshvar, Matthew Brandenburg, Christina Signoretti, Amy Zinski, Edward-James Gardner, Kelvin L Zheng, Chiderah P Abani, Carla Hu, Cameron P Beaudreault, Xiao-Lei Zhang, Patric K Stanton, Jun-Hyeong Cho, Libor Velíšek, Jana Velíšková, Saqlain Javed, Christopher S Leonard, Hae-Young Kim, Sangmi Chung
ABSTRACT
Previously, we demonstrated the efficacy of human pluripotent stem cell (hPSC)-derived GABAergic cortical interneuron (cIN) grafts in ameliorating seizures. However, a safe and reliable clinical translation requires a mechanistic understanding of graft function, as well as the assurance of long-term efficacy and safety. By employing hPSC-derived chemically matured migratory cINs in two models of epilepsy, we demonstrate lasting efficacy in treating seizures and comorbid deficits, as well as safety without uncontrolled growth. Host inhibition does not increase with increasing grafted cIN densities, assuring their safety without the risk of over-inhibition. Furthermore, their closed-loop optogenetic activation aborted seizure activity, revealing mechanisms of graft-mediated seizure control and allowing graft modulation for optimal translation. Monosynaptic tracing shows their extensive and specific synaptic connections with host neurons, resembling developmental connection specificity. These results offer confidence in stem cell-based therapy for epilepsy as a safe and reliable treatment for patients suffering from intractable epilepsy.
Sigma-Aldrich
DAPT, ≥98% (HPLC), solid
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-Choline Acetyltransferase Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Anti-Nuclei Antibody, clone 235-1, clone 235-1, Chemicon®, from mouse
Sigma-Aldrich
Anti-CaM Kinase II Antibody, α subunit, clone 6G9, clone 6G9, Upstate®, from mouse