Merck
  • NAD depletion mediates cytotoxicity in human neurons with autophagy deficiency.
NAD depletion mediates cytotoxicity in human neurons with autophagy deficiency.
Cell reports (2023-04-22)
Congxin Sun, Elena Seranova, Malkiel A Cohen, Miruna Chipara, Jennie Roberts, Dewi Astuti, Adina M Palhegyi, Animesh Acharjee, Lucia Sedlackova, Tetsushi Kataura, Elsje G Otten, Prashanta K Panda, Samuel Lara-Reyna, Miriam E Korsgen, Kevin J Kauffman, Alejandro Huerta-Uribe, Malgorzata Zatyka, Luiz F S E Silva, Jorge Torresi, Shupei Zhang, Georgina W Hughes, Carl Ward, Erich R Kuechler, David Cartwright, Sergey Trushin, Eugenia Trushina, Gaurav Sahay, Yosef Buganim, Gareth G Lavery, Joerg Gsponer, Daniel G Anderson, Eva-Maria Frickel, Tatiana R Rosenstock, Timothy Barrett, Oliver D K Maddocks, Daniel A Tennant, Haoyi Wang, Rudolf Jaenisch, Viktor I Korolchuk, Sovan Sarkar
ABSTRACT
Autophagy is a homeostatic process critical for cellular survival, and its malfunction is implicated in human diseases including neurodegeneration. Loss of autophagy contributes to cytotoxicity and tissue degeneration, but the mechanistic understanding of this phenomenon remains elusive. Here, we generated autophagy-deficient (ATG5-/-) human embryonic stem cells (hESCs), from which we established a human neuronal platform to investigate how loss of autophagy affects neuronal survival. ATG5-/- neurons exhibit basal cytotoxicity accompanied by metabolic defects. Depletion of nicotinamide adenine dinucleotide (NAD) due to hyperactivation of NAD-consuming enzymes is found to trigger cell death via mitochondrial depolarization in ATG5-/- neurons. Boosting intracellular NAD levels improves cell viability by restoring mitochondrial bioenergetics and proteostasis in ATG5-/- neurons. Our findings elucidate a mechanistic link between autophagy deficiency and neuronal cell death that can be targeted for therapeutic interventions in neurodegenerative and lysosomal storage diseases associated with autophagic defect.
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Rabbit Anti-Goat IgG, H & L Chain Specific Peroxidase Conjugate, liquid, Calbiochem®
Sigma-Aldrich
Oligomycin, A mixture of A, B, and C isomers.
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Goat Anti-Rabbit IgG, H & L Chain Specific Peroxidase Conjugate, liquid, Calbiochem®
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture
Sigma-Aldrich
FK866 hydrochloride hydrate, ≥98% (HPLC)
Sigma-Aldrich
Monoclonal Anti-Tubulin, Acetylated antibody produced in mouse, clone 6-11B-1, ascites fluid
Sigma-Aldrich
Rotenone, ≥95%
Sigma-Aldrich
Antimycin A from Streptomyces sp.
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Goat Anti-Mouse IgG, H&L Chain Specific Peroxidase Conjugate, liquid, Calbiochem®