Merck
  • Diverse effects of prostacyclin on angiogenesis-related processes in the porcine endometrium.
Diverse effects of prostacyclin on angiogenesis-related processes in the porcine endometrium.
Scientific reports (2023-08-30)
Magdalena Szymanska, Agnieszka Blitek
ABSTRACT
Angiogenesis is important for endometrial remodeling in mature females. The endometrium synthesizes high amounts of prostacyclin (PGI2) but the role of PGI2 in angiogenesis-related events in this tissue was not fully described. In the present study, porcine endometrial endothelial (pEETH) cells and/or a swine umbilical vein endothelial cell line (G1410 cells) were used to determine the regulation of PGI2 synthesis and PGI2 receptor (PTGIR) expression by cytokines and to evaluate the effect of PGI2 on pro-angiogenic gene expression, intracellular signaling activation, cell proliferation and migration, cell cycle distribution, and capillary-like structure formation. We found that IL1β, IFNγ, and/or TNFα increased PGI2 secretion and PTGIR expression in pEETH cells. Iloprost (a PGI2 analogue) acting through PTGIR enhanced the transcript abundance of KDR, FGFR2, and ANGPT2 and increased proliferation of pEETH cells. This latter was mediated by PI3K and mTOR activation. In support, transfection of G1410 cells with siRNA targeting PGI2 synthase decreased pro-angiogenic gene expression and cell proliferation. Furthermore, iloprost accelerated the gap closure and promoted cell cycle progression. Intriguingly, the formation of capillary-like structures was inhibited but not completely blocked by iloprost. These findings point to a complex pleiotropic role of PGI2 in angiogenesis-related events in the porcine uterus.
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Sigma-Aldrich
SQ 22536, Cell-permeable adenylate cyclase inhibitor.
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution