Merck
  • TGS1 impacts snRNA 3'-end processing, ameliorates survival motor neuron-dependent neurological phenotypes in vivo and prevents neurodegeneration.
TGS1 impacts snRNA 3'-end processing, ameliorates survival motor neuron-dependent neurological phenotypes in vivo and prevents neurodegeneration.
Nucleic acids research (2022-08-11)
Lu Chen, Caitlin M Roake, Paolo Maccallini, Francesca Bavasso, Roozbeh Dehghannasiri, Pamela Santonicola, Natalia Mendoza-Ferreira, Livia Scatolini, Ludovico Rizzuti, Alessandro Esposito, Ivan Gallotta, Sofia Francia, Stefano Cacchione, Alessandra Galati, Valeria Palumbo, Marie A Kobin, Gian Gaetano Tartaglia, Alessio Colantoni, Gabriele Proietti, Yunming Wu, Matthias Hammerschmidt, Cristiano De Pittà, Gabriele Sales, Julia Salzman, Livio Pellizzoni, Brunhilde Wirth, Elia Di Schiavi, Maurizio Gatti, Steven E Artandi, Grazia D Raffa
ABSTRACT
Trimethylguanosine synthase 1 (TGS1) is a highly conserved enzyme that converts the 5'-monomethylguanosine cap of small nuclear RNAs (snRNAs) to a trimethylguanosine cap. Here, we show that loss of TGS1 in Caenorhabditis elegans, Drosophila melanogaster and Danio rerio results in neurological phenotypes similar to those caused by survival motor neuron (SMN) deficiency. Importantly, expression of human TGS1 ameliorates the SMN-dependent neurological phenotypes in both flies and worms, revealing that TGS1 can partly counteract the effects of SMN deficiency. TGS1 loss in HeLa cells leads to the accumulation of immature U2 and U4atac snRNAs with long 3' tails that are often uridylated. snRNAs with defective 3' terminations also accumulate in Drosophila Tgs1 mutants. Consistent with defective snRNA maturation, TGS1 and SMN mutant cells also exhibit partially overlapping transcriptome alterations that include aberrantly spliced and readthrough transcripts. Together, these results identify a neuroprotective function for TGS1 and reinforce the view that defective snRNA maturation affects neuronal viability and function.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Anti-SMN Antibody, clone 2B1, clone 2B1, from mouse