Merck
A Bioluminescent Probe for Detecting Norepinephrine in Vivo.
Analytical chemistry (2022-04-23)
Zeping Zuo, Ting Kang, Shilong Hu, Wuyue Su, Yu Gan, Zhuang Miao, Hanqing Zhao, Ping Feng, Bowen Ke, Minyong Li
ABSTRACT
As a neurotransmitter, norepinephrine (NE) is critical for psychiatric conditions, neurodegenerative diseases, and pheochromocytoma. A real-time and noninvasive method for the detection of NE as a tracer to investigate the NE-relevant disease treatment process is urgently desirable. Herein, we successfully developed a turn-on NE bioluminescent probe (NBP), which was grounded on p-toluenethiol deprotectrf by nucleophilic substitution. Compared with other analytes, the NBP exhibited high sensitivity and selectivity in vitro. More importantly, the NBP provides a promising strategy for in vivo imaging of NE in living animals with noninvasive visualization and real-time features.
Sigma-Aldrich
NBP, ≥95% (HPLC)