Merck
  • Targeting ribosomal G-quadruplexes with naphthalene-diimides as RNA polymerase I inhibitors for colorectal cancer treatment.
Targeting ribosomal G-quadruplexes with naphthalene-diimides as RNA polymerase I inhibitors for colorectal cancer treatment.
Cell chemical biology (2021-06-25)
Victoria Sanchez-Martin, David A Schneider, Matilde Ortiz-Gonzalez, Ana Soriano-Lerma, Angel Linde-Rodriguez, Virginia Perez-Carrasco, Jose Gutierrez-Fernandez, Marta Cuadros, Carlos González, Miguel Soriano, Jose A Garcia-Salcedo
ABSTRACT
Guanine quadruplexes (G4s) are non-canonical nucleic acid structures commonly found in regulatory genomic regions. G4 targeting has emerged as a therapeutic approach in cancer. We have screened naphthalene-diimides (NDIs), a class of G4 ligands, in a cellular model of colorectal cancer (CRC). Here, we identify the leading compound T5 with a potent and selective inhibition of cell growth by high-affinity binding to G4s in ribosomal DNA, impairing RNA polymerase I (Pol I) elongation. Consequently, T5 induces a rapid inhibition of Pol I transcription, nucleolus disruption, proteasome-dependent Pol I catalytic subunit A degradation and autophagy. Moreover, we attribute the higher selectivity of carbohydrate-conjugated T5 for tumoral cells to its preferential uptake through the overexpressed glucose transporter 1. Finally, we succinctly demonstrate that T5 could be explored as a therapeutic agent in a patient cohort with CRC. Therefore, we report a mode of action for these NDIs involving ribosomal G4 targeting.
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)