Merck
  • Identification of ARMH4 and WIPF3 as human podocyte proteins with potential roles in immunomodulation and cytoskeletal dynamics.
Identification of ARMH4 and WIPF3 as human podocyte proteins with potential roles in immunomodulation and cytoskeletal dynamics.
PloS one (2023-01-18)
Francesco De Luca, Michelle Kha, Karl Swärd, Martin E Johansson
ABSTRACT
The podocyte is a specialized cell type critically involved in maintaining the selective filtration barrier of the kidney. Podocytes are primary or secondary targets for a multitude of kidney diseases. Despite intense investigation, the transcriptome and proteome of human podocytes remain incompletely characterized. Here, we analyzed publicly available RNA-Seq data from human kidneys (n = 85) to computationally identify potential novel podocyte markers. For confirmation, we used an online histology resource followed by in-house staining of human kidneys and biochemical fractionation of glomeruli. Initial characterization of the novel podocyte transcripts was performed using viral overexpression and mRNA silencing. Several previously unrecognized gene products were identified that correlated to established podocyte markers on the RNA level and that were histologically localized to podocytes. ARMH4 (a.k.a. UT2 or C14orf37) and WIPF3 (a.k.a CR16) were among the hits. We show that these transcripts increase in response to overexpression of the podocyte transcription factor LMX1B. Overexpression of ARMH4 from low endogenous levels in primary kidney epithelial cells reduced the release of the inflammatory mediators IL-1B and IL-8 (CXCL8). The opposite effect was seen in mature human podocytes when ARMH4 was silenced. Overexpression of WIPF3 stabilized N-WASP, known to be required for maintenance of podocyte foot processes, and increased cell motility as shown using a scratch assay. Moreover, data from normal and diseased human kidneys showed that ARMH4 was downregulated in glomerular pathologies, while WIPF3 remained constantly expressed. ARMH4 and WIPF3 are new potential markers of human podocytes, where they may modulate inflammatory insults by controlling cytokine release and contribute to cytoskeletal dynamics, respectively.
Sigma-Aldrich
Collagenase from Clostridium histolyticum, for general use, Type I, ≥125 CDU/mg solid
Sigma-Aldrich
Anti-C14orf37 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-C14orf37 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody
Sigma-Aldrich
Anti-WIPF3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Standardized vial containing 2,000 Kunitz units of DNase I (D4527), vial of ≥0.25 mg total protein
Sigma-Aldrich
Anti-Podocin, Mouse monoclonal, clone PD-65, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse