Merck
  • ULK1-mediated phosphorylation regulates the conserved role of YKT6 in autophagy.
ULK1-mediated phosphorylation regulates the conserved role of YKT6 in autophagy.
Journal of cell science (2023-01-17)
Pablo Sánchez-Martín, Franziska Kriegenburg, Ludovico Alves, Julius Adam, Jana Elsaesser, Riccardo Babic, Hector Mancilla, Mariya Licheva, Georg Tascher, Christian Münch, Stefan Eimer, Claudine Kraft
ABSTRACT
Autophagy is a catabolic process during which cytosolic material is enwrapped in a newly formed double-membrane structure called the autophagosome, and subsequently targeted for degradation in the lytic compartment of the cell. The fusion of autophagosomes with the lytic compartment is a tightly regulated step and involves membrane-bound SNARE proteins. These play a crucial role as they promote lipid mixing and fusion of the opposing membranes. Among the SNARE proteins implicated in autophagy, the essential SNARE protein YKT6 is the only SNARE protein that is evolutionarily conserved from yeast to humans. Here, we show that alterations in YKT6 function, in both mammalian cells and nematodes, produce early and late autophagy defects that result in reduced survival. Moreover, mammalian autophagosomal YKT6 is phospho-regulated by the ULK1 kinase, preventing premature bundling with the lysosomal SNARE proteins and thereby inhibiting autophagosome-lysosome fusion. Together, our findings reveal that timely regulation of the YKT6 phosphorylation status is crucial throughout autophagy progression and cell survival.
Sigma-Aldrich
Wortmannin, from Penicillium funiculosum, ≥98% (HPLC and TLC)
Sigma-Aldrich
ULK Active human, recombinant, expressed in FreeStyle 293-F cells, ≥40% (SDS-PAGE)
Sigma-Aldrich
Anti-OPTN antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Antimycin A from Streptomyces sp.
Millipore
Monoclonal Anti-HA−Agarose antibody produced in mouse, clone HA-7, purified immunoglobulin, PBS suspension