Merck
  • Generation of dorsoventral human spinal cord organoids via functionalizing composite scaffold for drug testing.
Generation of dorsoventral human spinal cord organoids via functionalizing composite scaffold for drug testing.
iScience (2023-01-18)
Weiwei Xue, Bo Li, Huihui Liu, Yujie Xiao, Bo Li, Lei Ren, Huijuan Li, Zhicheng Shao
ABSTRACT
The spinal cord possesses highly complex, finely organized cytoarchitecture guided by two dorsoventral morphogenic organizing centers. Thus, generation of human spinal cord tissue in vitro is challenging. Here, we demonstrated a novel method for generation of human dorsoventral spinal cord organoids using composite scaffolds. Specifically, the spinal cord ventralizing signaling Shh agonist (SAG) was loaded into a porous chitosan microsphere (PCSM), then thermosensitive Matrigel was coated on the surface to form composite microspheres with functional sustained-release SAG, termed as PCSM-Matrigel@SAG. Using PCSM-Matrigel@SAG as the core to induce 3D engineering of human spinal cord organoids from human pluripotent stem cells (ehSC-organoids), we found ehSC-organoids could form dorsoventral spinal cord-like cytoarchitecture with major domain-specific progenitors and neurons. Besides, these ehSC-organoids also showed functional calcium activity. In summary, these ehSC-organoids are of great significance for modeling spinal cord development, drug screening as 3D models for motor neuron diseases, and spinal cord injury repair.
Sigma-Aldrich
holo-Transferrin human, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Insulin human, recombinant, expressed in yeast (proprietary host)
Sigma-Aldrich
Anti-Human Leukemia Inhibitory Factor Antibody, clone 4F7.2, clone 4F7.2, Chemicon®, from mouse
Sigma-Aldrich
Anti-GAD67 Antibody, clone 1G10.2, clone 1G10.2, Chemicon®, from mouse
Sigma-Aldrich
Anti-Stage-Specific Embryonic Antigen-4 Antibody, clone MC-813-70, clone MC-813-70, Chemicon®, from mouse
Sigma-Aldrich
Anti-Choline Acetyltransferase Antibody, Chemicon®, from goat
Roche
DNase I, from bovine pancreas
Sigma-Aldrich
Anti-Olig-2 Antibody, Chemicon®, from rabbit