Merck
  • Phosphorylcholine Monoclonal Antibody Therapy Decreases Intraplaque Angiogenesis and Intraplaque Hemorrhage in Murine Vein Grafts.
Phosphorylcholine Monoclonal Antibody Therapy Decreases Intraplaque Angiogenesis and Intraplaque Hemorrhage in Murine Vein Grafts.
International journal of molecular sciences (2022-11-12)
Fabiana Baganha, Thijs J Sluiter, Rob C M de Jong, Louise A van Alst, Hendrika A B Peters, J Wouter Jukema, Mirela Delibegovic, Knut Pettersson, Paul H A Quax, Margreet R de Vries
ABSTRACT
Phosphorylcholine (PC) is one of the main oxLDL epitopes playing a central role in atherosclerosis, due to its atherogenic and proinflammatory effects. PC can be cleared by natural IgM antibodies and low levels of these antibodies have been associated with human vein graft (VG) failure. Although PC antibodies are recognized for their anti-inflammatory properties, their effect on intraplaque angiogenesis (IPA) and intraplaque hemorrhage (IPH)-interdependent processes contributing to plaque rupture-are unknown. We hypothesized that new IgG phosphorylcholine antibodies (PC-mAb) could decrease vulnerable lesions in murine VGs.Therefore, hypercholesterolemic male ApoE3*Leiden mice received a (donor) caval vein interposition in the carotid artery and weekly IP injections of (5 mg/kg) PCmAb (n = 11) or vehicle (n = 12) until sacrifice at day 28. We found that PCmAb significantly decreased vein graft media (13%), intima lesion (25%), and increased lumen with 32% compared to controls. PCmAb increased collagen content (18%) and decreased macrophages presence (31%). PCmAb resulted in 23% decreased CD163+ macrophages content in vein grafts whereas CD163 expression was decreased in Hb:Hp macrophages. PCmAb significantly lowered neovessel density (34%), EC proliferation and migration with/out oxLDL stimulation. Moreover, PCmAb enhanced intraplaque angiogenic vessels maturation by increasing neovessel pericyte coverage in vivo (31%). Together, this resulted in a 62% decrease in IPH. PCmAb effectively inhibits murine atherosclerotic lesion formation in vein grafts by reducing IPA and IPH via decreased neovessel density and macrophages influx and increased neovessel maturation. PC-mAb therefore holds promise as a new therapeutic approach to prevent vein graft disease.
Sigma-Aldrich
Haptoglobin (Phenotype 1-1) from human plasma, ≥95% (SDS-PAGE)
Sigma-Aldrich
Hemoglobin A0, Ferrous Stabilized human, lyophilized powder
Roche
Collagen, from rat tail tendon
Sigma-Aldrich
Trypsin-EDTA solution, 0.25%, sterile-filtered, BioReagent, suitable for cell culture, 2.5 g porcine trypsin and 0.2 g EDTA, 4Na per liter of Hanks′ Balanced Salt Solution with phenol red