Merck
  • MITF Is Regulated by Redox Signals Controlled by the Selenoprotein Thioredoxin Reductase 1.
MITF Is Regulated by Redox Signals Controlled by the Selenoprotein Thioredoxin Reductase 1.
Cancers (2022-10-28)
Chelsey D Kline, Madeleine Anderson, John W Bassett, Gail Kent, Rachel Berryman, Matthew Honeggar, Shosuke Ito, Kazumasa Wakamatsu, Arup K Indra, Philip J Moos, Sancy A Leachman, Pamela B Cassidy
ABSTRACT
TR1 and other selenoproteins have paradoxical effects in melanocytes and melanomas. Increasing selenoprotein activity with supplemental selenium in a mouse model of UV-induced melanoma prevents oxidative damage to melanocytes and delays melanoma tumor formation. However, TR1 itself is positively associated with progression in human melanomas and facilitates metastasis in melanoma xenografts. Here, we report that melanocytes expressing a microRNA directed against TR1 (TR1low) grow more slowly than control cell lines and contain significantly less melanin. This phenotype is associated with lower tyrosinase (TYR) activity and reduced transcription of tyrosinase-like protein-1 (TYRP1). Melanoma cells in which the TR1 gene (TXNRD1) was disrupted using Crispr/Cas9 showed more dramatic effects including the complete loss of the melanocyte-specific isoform of MITF; other MITF isoforms were unaffected. We provide evidence that TR1 depletion results in oxidation of MITF itself. This newly discovered mechanism for redox modification of MITF has profound implications for controlling both pigmentation and tumorigenesis in cells of the melanocyte lineage.
Sigma-Aldrich
Anti-TXNRD1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
hBFGF, FGF-Basic, recombinant, expressed in E. coli, suitable for cell culture
SAFC
Insulin, Human Recombinant, for research or for further manufacturing use, dry powder
Sigma-Aldrich
Anti-TXN antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Microphthalmia (Mi) Antibody, clone C5, clone C5, Chemicon®, from mouse