Merck
  • Chondroitin sulfate proteoglycan is a potential target of memantine to improve cognitive function via the promotion of adult neurogenesis.
Chondroitin sulfate proteoglycan is a potential target of memantine to improve cognitive function via the promotion of adult neurogenesis.
British journal of pharmacology (2022-07-08)
Shoichiro Maeda, Jun Yamada, Kyoko M Iinuma, Satomi Nadanaka, Hiroshi Kitagawa, Shozo Jinno
ABSTRACT
Chondroitin sulfate proteoglycan (CSPG) constitutes the neurogenic niche in the hippocampus. The reduction of hippocampal neurogenesis is involved in ageing-related cognitive decline and dementia. The purpose of this study is to find candidates that improve cognitive function by analysing the effects of memantine (MEM), a therapeutic agent for Alzheimer's disease, on CSPG and adult hippocampal neurogenesis. The effects of MEM on neurogenesis-related cells and CSPG content were assessed in the hippocampus of middle-aged mice. The MEM-induced alterations in gene expressions of neurotrophins and enzymes associated with biosynthesis and degradation of CSPG in the hippocampus also were measured. The effects of MEM on cognitive function were estimated using a behavioural test battery. The same set of behavioural tests was applied to evaluate the effects of pharmacological depletion of CSPG in the hippocampus. The densities of newborn granule cells and content of CSPG in the hippocampus were increased by MEM. The expression levels of the enzyme responsible for the biosynthesis CSPG were increased by MEM. The neurotrophin-related molecules were activated by MEM. Short- and long-term memory performance was improved by MEM. Pharmacological depletion of CSPG impairs the effects of MEM on cognitive improvement in middle-aged mice. MEM regulates the biosynthesis and degradation of CSPG, which may underlie the improvement of cognitive function via the promotion of adult hippocampal neurogenesis. These results imply that CSPG-related enzymes potentially may be attractive candidates for the treatment of ageing-related cognitive decline.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-Heparan Sulfate Proteoglycan (Perlecan) Antibody, clone 5D7-2E4, clone 5D7-2E4, from mouse
Sigma-Aldrich
Monoclonal Anti-Phosphacan antibody produced in mouse, ~2 mg/mL, clone 122.2, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-Aggrecan Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Keratan Sulfate Antibody, clone EFG-11, ascites fluid, clone EFG-11, Chemicon®
Sigma-Aldrich
Anti-Neurocan Antibody, clone 650.24, clone 650.24, Chemicon®, from mouse