Merck
  • Inhibition of cysteine cathepsin B and L activation in astrocytes contributes to neuroprotection against cerebral ischemia via blocking the tBid-mitochondrial apoptotic signaling pathway.
Inhibition of cysteine cathepsin B and L activation in astrocytes contributes to neuroprotection against cerebral ischemia via blocking the tBid-mitochondrial apoptotic signaling pathway.
Glia (2014-03-13)
Min Xu, Lei Yang, Jia-Guo Rong, Yong Ni, Wei-Wei Gu, Yu Luo, Kazumi Ishidoh, Nobuhiko Katunuma, Zhong-Sheng Li, Hui-Ling Zhang
ABSTRACT
The roles of cathepsins in the ischemic astrocytic injury remain unclear. Here, we test the hypothesis that activation of cathepsin B and L contributes to the ischemic astrocyte injury via the tBid-mitochondrial apoptotic signaling pathways. In the rat models of pMCAO, CA-074Me or Clik148, a selective inhibitor of cathepsin B or cathepsin L, reduced the infarct volume, improved the neurological deficits and increased the MAP2 and GFAP levels. In OGD-induced astrocyte injury, CA-074Me or Clik148 decreased the LDH leakage and increased the GFAP levels. In the ischemic cortex or OGD-induced astrocytes injury, Clik148 or CA-074Me reversed pMCAO or OGD-induced increase in active cathepsin L or cathepsin B at 3 h or 6 h, increase in tBid, reduction in mitochondrial cytochrome-c (Cyt-c) and increase in cytoplastic Cyt-c and active caspase-3 at 12-24 h of the late stage of pMCAO or OGD. CA-074Me or Clik148 also reduced cytosolic and mitochondrial tBid, increased mitochondrial Cyt-c and decreased cytoplastic Cyt-c and active caspase-3 at 6 h of the early stage of Bid activation. CA-074Me or Clik148 blocked the pMCAO-induced release of cathepsin B or L from the lysosomes into the cytoplasm and activation of caspase-3 in ischemic astrocytes at 12 h after ischemia. Concurrent inhibition of cathepsin B and cathepsin L provided better protection on the OGD-induced astrocytic apoptosis than obtained with separate use of each inhibitor. These results suggest that inhibition of the cysteine cathepsin B and cathepsin L activation in ischemic astrocytes contributes to neuroprotection via blocking the tBid-mitochondrial apoptotic signaling pathway.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Monoclonal Anti-MAP2 antibody produced in mouse, clone HM-2, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein (GFAP) Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5, ascites fluid, clone GA5, Chemicon®
Sigma-Aldrich
Anti-Caspase 3 Antibody, active (cleaved) form, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Bid Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein (GFAP)−Cy3 antibody, Mouse monoclonal, clone G-A-5, purified from hybridoma cell culture