Merck
  • Assessment of mitochondrial respiratory chain enzymatic activities on tissues and cultured cells.
Assessment of mitochondrial respiratory chain enzymatic activities on tissues and cultured cells.
Nature protocols (2012-06-02)
Marco Spinazzi, Alberto Casarin, Vanessa Pertegato, Leonardo Salviati, Corrado Angelini
ABSTRACT
The assessment of mitochondrial respiratory chain (RC) enzymatic activities is essential for investigating mitochondrial function in several situations, including mitochondrial disorders, diabetes, cancer, aging and neurodegeneration, as well as for many toxicological assays. Muscle is the most commonly analyzed tissue because of its high metabolic rates and accessibility, although other tissues and cultured cell lines can be used. We describe a step-by-step protocol for a simple and reliable assessment of the RC enzymatic function (complexes I-IV) for minute quantities of muscle, cultured cells and isolated mitochondria from a variety of species and tissues, by using a single-wavelength spectrophotometer. An efficient tissue disruption and the choice for each assay of specific buffers, substrates, adjuvants and detergents in a narrow concentration range allow maximal sensitivity, specificity and linearity of the kinetics. This protocol can be completed in 3 h.
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, 98.5-101.5%, BioUltra
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Alconox® detergent, 0.5 oz packs
Sigma-Aldrich
Cytochrome c from equine heart, ≥95% based on Mol. Wt. 12,384 basis
Sigma-Aldrich
Antimycin A from Streptomyces sp.
Sigma-Aldrich
Cytochrome c from equine heart, ≥95% (SDS-PAGE)
Sigma-Aldrich
Rotenone, ≥95%
Sigma-Aldrich
PIP5-Kinase HTRF Assay, The PIP5 Kinase HTRF Assay kit provides a method for assaying PI(4)P-5 kinases in a homogenous 384 well format. Suitable for screening of potential inhibitors of these enzymes.