Merck
  • Reactive astrocytes acquire neuroprotective as well as deleterious signatures in response to Tau and Aß pathology.
Reactive astrocytes acquire neuroprotective as well as deleterious signatures in response to Tau and Aß pathology.
Nature communications (2022-01-12)
Zoeb Jiwaji, Sachin S Tiwari, Rolando X Avilés-Reyes, Monique Hooley, David Hampton, Megan Torvell, Delinda A Johnson, Jamie McQueen, Paul Baxter, Kayalvizhi Sabari-Sankar, Jing Qiu, Xin He, Jill Fowler, James Febery, Jenna Gregory, Jamie Rose, Jane Tulloch, Jamie Loan, David Story, Karina McDade, Amy M Smith, Peta Greer, Matthew Ball, Peter C Kind, Paul M Matthews, Colin Smith, Owen Dando, Tara L Spires-Jones, Jeffrey A Johnson, Siddharthan Chandran, Giles E Hardingham
ABSTRACT
Alzheimer's disease (AD) alters astrocytes, but the effect of Aß and Tau pathology is poorly understood. TRAP-seq translatome analysis of astrocytes in APP/PS1 ß-amyloidopathy and MAPTP301S tauopathy mice revealed that only Aß influenced expression of AD risk genes, but both pathologies precociously induced age-dependent changes, and had distinct but overlapping signatures found in human post-mortem AD astrocytes. Both Aß and Tau pathology induced an astrocyte signature involving repression of bioenergetic and translation machinery, and induction of inflammation pathways plus protein degradation/proteostasis genes, the latter enriched in targets of inflammatory mediator Spi1 and stress-activated cytoprotective Nrf2. Astrocyte-specific Nrf2 expression induced a reactive phenotype which recapitulated elements of this proteostasis signature, reduced Aß deposition and phospho-tau accumulation in their respective models, and rescued brain-wide transcriptional deregulation, cellular pathology, neurodegeneration and behavioural/cognitive deficits. Thus, Aß and Tau induce overlapping astrocyte profiles associated with both deleterious and adaptive-protective signals, the latter of which can slow patho-progression.
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Monoclonal Anti-Neurofilament 200 antibody produced in mouse, clone NE14, ascites fluid
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5, clone GA5, Chemicon®, from mouse
Sigma-Aldrich
Anti-Neurofilament 200 kDa Antibody, clone N52, clone N52, Chemicon®, from mouse