Merck
  • The 3' Pol II pausing at replication-dependent histone genes is regulated by Mediator through Cajal bodies' association with histone locus bodies.
The 3' Pol II pausing at replication-dependent histone genes is regulated by Mediator through Cajal bodies' association with histone locus bodies.
Nature communications (2022-05-26)
Hidefumi Suzuki, Ryota Abe, Miho Shimada, Tomonori Hirose, Hiroko Hirose, Keisuke Noguchi, Yoko Ike, Nanami Yasui, Kazuki Furugori, Yuki Yamaguchi, Atsushi Toyoda, Yutaka Suzuki, Tatsuro Yamamoto, Noriko Saitoh, Shigeo Sato, Chieri Tomomori-Sato, Ronald C Conaway, Joan W Conaway, Hidehisa Takahashi
ABSTRACT
Non-polyadenylated mRNAs of replication-dependent histones (RDHs) are synthesized by RNA polymerase II (Pol II) at histone locus bodies (HLBs). HLBs frequently associate with Cajal bodies (CBs), in which 3'-end processing factors for RDH genes are enriched; however, this association's role in transcription termination of RDH genes remains unclear. Here, we show that Pol II pauses immediately upstream of transcript end sites of RDH genes and Mediator plays a role in this Pol II pausing through CBs' association with HLBs. Disruption of the Mediator docking site for Little elongation complex (LEC)-Cap binding complex (CBC)-Negative elongation factor (NELF), components of CBs, interferes with CBs' association with HLBs and 3' Pol II pausing, resulting in increased aberrant unprocessed RDH gene transcripts. Our findings suggest Mediator's involvement in CBs' association with HLBs to facilitate 3' Pol II pausing and subsequent 3'-end processing of RDH genes by supplying 3'-end processing factors.
Sigma-Aldrich
Benzonase® Nuclease, ultrapure, ≥250 units/μL, ≥99% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous glycerol solution, ultrapure grade
Sigma-Aldrich
Anti-CDK11A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody
Sigma-Aldrich
Anti-LSM11 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Proteinase K from Tritirachium album, lyophilized powder, BioUltra, ≥30 units/mg protein, for molecular biology
Sigma-Aldrich
Anti-WRAP53 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody
Sigma-Aldrich
Anti-NPAT antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody
Sigma-Aldrich
Anti-CASP8AP2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody