Merck
  • PARP inhibition impedes the maturation of nascent DNA strands during DNA replication.
PARP inhibition impedes the maturation of nascent DNA strands during DNA replication.
Nature structural & molecular biology (2022-03-26)
Alina Vaitsiankova, Kamila Burdova, Margarita Sobol, Amit Gautam, Oldrich Benada, Hana Hanzlikova, Keith W Caldecott
ABSTRACT
Poly(ADP-ribose) polymerase 1 (PARP1) is implicated in the detection and processing of unligated Okazaki fragments and other DNA replication intermediates, highlighting such structures as potential sources of genome breakage induced by PARP inhibition. Here, we show that PARP1 activity is greatly elevated in chicken and human S phase cells in which FEN1 nuclease is genetically deleted and is highest behind DNA replication forks. PARP inhibitor reduces the integrity of nascent DNA strands in both wild-type chicken and human cells during DNA replication, and does so in FEN1-/- cells to an even greater extent that can be detected as postreplicative single-strand nicks or gaps. Collectively, these data show that PARP inhibitors impede the maturation of nascent DNA strands during DNA replication, and implicate unligated Okazaki fragments and other nascent strand discontinuities in the cytotoxicity of these compounds.
Sigma-Aldrich
Anti-DNA Antibody, single stranded, clone 16-19, clone 16-19, Chemicon®, from mouse
Sigma-Aldrich
Rabbit anti-Phospho RPA32 (S4/S8) Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.