Merck
  • Dynamic O-GlcNAcylation coordinates ferritinophagy and mitophagy to activate ferroptosis.
Dynamic O-GlcNAcylation coordinates ferritinophagy and mitophagy to activate ferroptosis.
Cell discovery (2022-05-04)
Fan Yu, Qianping Zhang, Hanyu Liu, Jinming Liu, Song Yang, Xiaofan Luo, Wei Liu, Hao Zheng, Qiqi Liu, Yunxi Cui, Guo Chen, Yanjun Li, Xinglu Huang, Xiyun Yan, Jun Zhou, Quan Chen
ABSTRACT
Ferroptosis is a regulated iron-dependent cell death characterized by the accumulation of lipid peroxidation. A myriad of facets linking amino acid, lipid, redox, and iron metabolisms were found to drive or to suppress the execution of ferroptosis. However, how the cells decipher the diverse pro-ferroptotic stress to activate ferroptosis remains elusive. Here, we report that protein O-GlcNAcylation, the primary nutrient sensor of glucose flux, orchestrates both ferritinophagy and mitophagy for ferroptosis. Following the treatment of ferroptosis stimuli such as RSL3, a commonly used ferroptosis inducer, there exists a biphasic change of protein O-GlcNAcylation to modulate ferroptosis. Pharmacological or genetic inhibition of O-GlcNAcylation promoted ferritinophagy, resulting in the accumulation of labile iron towards mitochondria. Inhibition of O-GlcNAcylation resulted in mitochondria fragmentation and enhanced mitophagy, providing an additional source of labile iron and rendering the cell more sensitive to ferroptosis. Mechanistically, we found that de-O-GlcNAcylation of the ferritin heavy chain at S179 promoted its interaction with NCOA4, the ferritinophagy receptor, thereby accumulating labile iron for ferroptosis. Our findings reveal a previously uncharacterized link of dynamic O-GlcNAcylation with iron metabolism and decision-making for ferroptosis, thus offering potential therapeutic intervention for fighting disease.
Sigma-Aldrich
Monoclonal Anti-O-GlcNAc antibody produced in mouse, clone 9H6
Sigma-Aldrich
Anti-c-Myc antibody, Mouse monoclonal, clone 9E10, purified from hybridoma cell culture
Sigma-Aldrich
Anti-β-O-Linked N-Acetylglucosamine antibody, Mouse monoclonal, clone CTD110.6, purified from hybridoma cell culture
Roche
Anti-GFP, from mouse IgG1κ (clones 7.1 and 13.1)