Merck
  • Hydrogen sulfide-induced GAPDH sulfhydration disrupts the CCAR2-SIRT1 interaction to initiate autophagy.
Hydrogen sulfide-induced GAPDH sulfhydration disrupts the CCAR2-SIRT1 interaction to initiate autophagy.
Autophagy (2021-01-19)
Iram Khan Iqbal, Sapna Bajeli, Shivani Sahu, Shabir Ahmad Bhat, Ashwani Kumar
ABSTRACT
The deacetylase SIRT1 (sirtuin 1) has emerged as a major regulator of nucleocytoplasmic distribution of macroautophagy/autophagy marker MAP1LC3/LC3 (microtubule-associated protein 1 light chain 3). Activation of SIRT1 leads to the deacetylation of LC3 and its translocation from the nucleus into the cytoplasm leading to an increase in the autophagy flux. Notably, hydrogen sulfide (H2S) is a cytoprotective gasotransmitter known to activate SIRT1 and autophagy; however, the underlying mechanism for both remains unknown. Herein, we demonstrate that H2S sulfhydrates the active site cysteine of the glycolytic enzyme GAPDH (glyceraldehyde-3-phosphate dehydrogenase). Sulfhydration of GAPDH leads to its redistribution into the nucleus. Importantly, nuclear localization of GAPDH is critical for H2S-mediated activation of autophagy as H2S does not induce autophagy in cells with GAPDH ablation or cells overexpressing a GAPDH mutant lacking the active site cysteine. Importantly, we observed that nuclear GAPDH interacts with CCAR2/DBC1 (cell cycle activator a nd apoptosis regulator 2) inside the nucleus. CCAR2 interacts with the deacetylase SIRT1 to inhibit its activity. Interaction of GAPDH with CCAR2 disrupts the inhibitory effect of CCAR2 on SIRT1. Activated SIRT1 then deacetylates MAP1LC3B/LC3B (microtubule-associated protein 1 light chain 3 beta) to induce its translocation into the cytoplasm and activate autophagy. Additionally, we demonstrate this pathway's physiological role in autophagy-mediated trafficking of Mycobacterium tuberculosis into lysosomes to restrict intracellular mycobacteria growth. We think that the pathway described here could be involved in H2S-mediated clearance of intracellular pathogens and other health benefits.Abbreviations: ATG5: autophagy related 5; ATG7: autophagy related 7; BECN1: beclin 1, autophagy related; CCAR2/DBC1: cell cycle activator and apoptosis regulator 2; CFU: colony-forming units; DLG4/PSD95: discs large MAGUK scaffold protein 4; EX-527: 6-chloro-2,3,4,9-tetrahydro-1H-carbazole-1-carboxamide; GAPDH: glyceraldehyde-3-phosphate dehydrogenase; H2S: hydrogen sulfide; HEK: human embryonic kidney cells; MAP1LC3B/LC3B: microtubule-associated protein 1 light chain 3 beta; MEF: mouse embryonic fibroblast; Mtb: Mycobacterium tuberculosis; MTOR: mechanistic target of rapamycin kinase; MOI: multiplicity of infection; NO: nitric oxide; PI3K: phosphatidylinositol-4,5-bisphosphate 3-kinase; PLA: proximity ligation assay; PRKAA: protein kinase, AMP-activated, alpha catalytic subunit; SIAH1: siah E3 ubiquitin protein ligase 1A; SIRT1: sirtuin 1; TB: tuberculosis; TP53INP2/DOR: transformation related protein 53 inducible nuclear protein 2; TRP53/TP53: transformation related protein 53.
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)ā€“Peroxidase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
BenzonaseĀ® Nuclease, ultrapure, ā‰„250 units/Ī¼L, ā‰„99% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous glycerol solution, ultrapure grade
Sigma-Aldrich
Wortmannin, Ready Made Solution, from Penicillium funiculosum, ā‰„95% (HPLC)
Sigma-Aldrich
Monoclonal ANTI-FLAGĀ® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
AMPK Inhibitor, Compound C, InSolution, ā‰„95%, 10 mM, AMPK Inhibitor
Sigma-Aldrich
EX-527, ≥98% (HPLC)
Millipore
EZviewā„¢ Red Streptavidin Affinity Gel
Millipore
ANTI-FLAGĀ® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Catalase from bovine liver, powder, suitable for cell culture, 2,000-5,000 units/mg protein
Sigma-Aldrich
DuolinkĀ® In Situ PLAĀ® Probe Anti-Mouse MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)
Sigma-Aldrich
Bafilomycin A1 from Streptomyces griseus, ā‰„90% (HPLC)
Sigma-Aldrich
Monoclonal Anti-Ī²-Actin antibody produced in mouse, clone AC-15, ascites fluid