Merck
  • Long non-coding RNA OIP5-AS1 promotes the progression of esophageal cancer by regulating miR-30a/VOPP1 expression.
Long non-coding RNA OIP5-AS1 promotes the progression of esophageal cancer by regulating miR-30a/VOPP1 expression.
Oncology letters (2021-08-14)
Jiajun Xu, Zhixi Chen, Zheng Fang, Shixiong Chen, Ying Guo, Xianfeng Liu, Kai Chen, Shengjia Chen
ABSTRACT
Long non-coding RNAs (lncRNAs) serve an important role in the development of esophageal cancer (EC), which is the eighth most common type of cancer worldwide. lncRNA opa-interacting protein 5 antisense transcript 1 (OIP5-AS1) is associated with human malignancy. However, the biological roles of OIP5-AS1 in the development of EC remain unclear. In the present study, transfection was conducted, and reverse transcription-quantitative PCR and western blot analysis were used for the detection of mRNA and protein expression, respectively. Furthermore, dual-luciferase reporter and RNA immunoprecipitation assays were used to study the interaction between miRNA and lncRNA or genes. The results revealed that OIP5-AS1 expression in EC tissues and cultured EC cells was upregulated, microRNA-30a (miR-30a) expression was downregulated. OIP5-AS1-knockdown suppressed the proliferation, migration and invasion of EC9706 and EC109 cells. miR-30a was confirmed to interact with OIP5-AS1, and miR-30a-mimics transfection ameliorated the effects of OIP5-AS1 in EC cells. Vesicular overexpressed in cancer prosurvival protein 1 (VOPP1) was verified as the direct target of miR-30a. VOPP1 expression was positively correlated with OIP5-AS1 expression in EC cells. Overexpression of VOPP1 ameliorated the negative effects of OIP5-AS1-knockdown on EC9706 and EC109 cells. In conclusion, OIP5-AS1 promoted the proliferation, migration and invasion of EC cells by increasing VOPP1 expression by sponging miR-30a.
Sigma-Aldrich
IgG from sheep serum, reagent grade, ≥95% (SDS-PAGE), essentially salt-free, lyophilized powder
Sigma-Aldrich
Anti-pan Ago Antibody, clone 2A8, clone 2A8, from mouse
Sigma-Aldrich
Anti-WBP1L antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-GAPDHS antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution