Merck
  • Temperature response of enriched pre-pubertal caprine male germline stem cells in vitro.
Temperature response of enriched pre-pubertal caprine male germline stem cells in vitro.
Cell stress & chaperones (2021-09-24)
Shiva P Singh, Suresh D Kharche, Manisha Pathak, Yogesh K Soni, Kumaresan Gururaj, Atul K Sharma, Manoj K Singh, Manmohan S Chauhan
ABSTRACT
The present study aims to evaluate culture temperature-dependent variation in survival, growth characteristics and expression of stress, pluripotency, apoptosis, and adhesion markers in enriched caprine male germline stem cells (cmGSCs). For this, testes from pre-pubertal bucks (4-5 months; n = 4) were used to isolated cells by a two-step enzymatic digestion method. After enrichment of cmGSCs by multiple methods (differential platting, Percoll density gradient centrifugation, and MACS), viability of CD90+ cells was assessed before co-cultured onto the Sertoli cell feeder layer at different temperatures (35.5, 37.0, 38.5, and 40.0 °C). The culture characteristics of cells were compared with MTT assay (viability); cluster-forming activity assay, SA-β1-gal assay (senescence), BrdU assay (proliferation), and transcript expression analyses by qRT-PCR. Moreover, the co-localization of pluripotency markers (UCHL-1, PLZF, and DBA) was examined by a double-immunofluorescence method. The cells grown at 37.0 °C showed faster proliferation with a significantly (p < 0.05) higher number of viable cells and greater number of cell clusters, besides higher expression of pluripotency markers. The transcript expression of HSPs (more noticeably HSP72 than HSP73), anti-oxidative enzymes (GPx and CuZnSOD), and adhesion molecule (β1-integrin) was significantly (p < 0.05) downregulated when grown at 35.0, 38.5, or 40.0 °C compared with 37.0 °C. The expression of pluripotency-specific transcripts was significantly (p < 0.05) lower in cmGSCs grown at the culture temperature lower (35.5 °C) or higher (38.5 °C and 40.0 °C) than 37.0 °C. Overall, the culture temperature significantly affects the proliferation, growth characteristics, and expression of heat stress, pluripotency, and adhesion-specific markers in pre-pubertal cmGSCs. These results provide an insight to develop strategies for the improved cultivation and downstream applications of cmGSCs.
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of physiologically active rat hepatocytes, Type IV, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
4′,6-Diamidino-2-phenylindole dihydrochloride, powder, BioReagent, suitable for cell culture, ≥98% (HPLC and TLC), suitable for fluorescence
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Milli-Mark® Anti-BrdU Antibody, clone BU-1 Alexa Fluor 488 conjugated, clone Bu-1, Milli-Mark®, from mouse
Sigma-Aldrich
Trypsin from porcine pancreas, lyophilized powder, Type II-S, 1,000-2,000 units/mg dry solid
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, lyophilized powder, Protein ≥85 %, ≥400 Kunitz units/mg protein
Sigma-Aldrich
Hyaluronidase from sheep testes, Type II, lyophilized powder, ≥300 units/mg solid
Sigma-Aldrich
MTT Cell Growth Assay Kit, MTT Cell Growth Assay is a colorimetric assay that can be used for either proliferation or complement-mediated cytotoxicity assays.