Merck
  • ATF-4 and hydrogen sulfide signalling mediate longevity in response to inhibition of translation or mTORC1.
ATF-4 and hydrogen sulfide signalling mediate longevity in response to inhibition of translation or mTORC1.
Nature communications (2022-02-20)
Cyril Statzer, Jin Meng, Richard Venz, Monet Bland, Stacey Robida-Stubbs, Krina Patel, Dunja Petrovic, Raffaella Emsley, Pengpeng Liu, Ianessa Morantte, Cole Haynes, William B Mair, Alban Longchamp, Milos R Filipovic, T Keith Blackwell, Collin Y Ewald
ABSTRACT
Inhibition of the master growth regulator mTORC1 (mechanistic target of rapamycin complex 1) slows ageing across phyla, in part by reducing protein synthesis. Various stresses globally suppress protein synthesis through the integrated stress response (ISR), resulting in preferential translation of the transcription factor ATF-4. Here we show in C. elegans that inhibition of translation or mTORC1 increases ATF-4 expression, and that ATF-4 mediates longevity under these conditions independently of ISR signalling. ATF-4 promotes longevity by activating canonical anti-ageing mechanisms, but also by elevating expression of the transsulfuration enzyme CTH-2 to increase hydrogen sulfide (H2S) production. This H2S boost increases protein persulfidation, a protective modification of redox-reactive cysteines. The ATF-4/CTH-2/H2S pathway also mediates longevity and increased stress resistance from mTORC1 suppression. Increasing H2S levels, or enhancing mechanisms that H2S influences through persulfidation, may represent promising strategies for mobilising therapeutic benefits of the ISR, translation suppression, or mTORC1 inhibition.
Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis
Sigma-Aldrich
α-Amanitin, from Amanita phalloides, ≥85% (HPLC), powder
Sigma-Aldrich
Tunicamycin from Streptomyces sp.
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Sodium fluoride 0.5 M solution
Roche
Anti-GFP, from mouse IgG1κ (clones 7.1 and 13.1)