Merck
  • Polymerase synthesis of four-base DNA from two stable dimeric nucleotides.
Polymerase synthesis of four-base DNA from two stable dimeric nucleotides.
Nucleic acids research (2019-09-11)
Michael G Mohsen, Debin Ji, Eric T Kool
ABSTRACT
We document the preparation and properties of dimerized pentaphosphate-bridged deoxynucleotides (dicaptides) that contain reactive components of two different nucleotides simultaneously. Importantly, dicaptides are found to be considerably more stable to hydrolysis than standard dNTPs. Steady-state kinetics studies show that the dimers exhibit reasonably good efficiency with the Klenow fragment of DNA polymerase I, and we identify thermostable enzymes that process them efficiently at high temperature. Experiments show that the dAp5dT dimer successfully acts as a combination of dATP and dTTP in primer extension reactions, and the dGp5dC dimer as a combination of dGTP and dCTP. The two dimers in combination promote successful 4-base primer extension. The final byproduct of the reaction, triphosphate, is shown to be less inhibitory to primer extension than pyrophosphate, the canonical byproduct. Finally, we document PCR amplification of DNA with two dimeric nucleotides, and show that the dimers can promote amplification under extended conditions when PCR with normal dNTPs fails. These dimeric nucleotides represent a novel and simple approach for increasing stability of nucleotides and avoiding inhibition from pyrophosphate.
Sigma-Aldrich
2′-Deoxyguanosine 5′-triphosphate trisodium salt solution, 10 mM in H2O
Sigma-Aldrich
2′-Deoxyadenosine 5′-monophosphate, Sigma Grade, 98-100%
Sigma-Aldrich
2′-Deoxyadenosine 5′-triphosphate sodium salt solution, 100 mM, pH 7
Sigma-Aldrich
Thymidine 5′-triphosphate sodium salt solution, 10 mM (pH 7.0)
Sigma-Aldrich
2′-Deoxycytidine 5′-monophosphate, Sigma Grade, ≥95.0%
Sigma-Aldrich
Deoxynucleotide Set, 100 mM, Individual dNTPs for routine PCR; 1 mL each
Sigma-Aldrich
Deoxynucleotide Set, 100 mM, Individual dNTPs for routine PCR; 0.25 mL each
Sigma-Aldrich
2′-Deoxycytidine 5′-triphosphate disodium salt, 100 mM (pH 7)