Merck
  • Highly sensitive electrochemiluminescence immunosensor based on ABEI/H2O2 system with PFO dots as enhancer for detection of kidney injury molecule-1.
Highly sensitive electrochemiluminescence immunosensor based on ABEI/H2O2 system with PFO dots as enhancer for detection of kidney injury molecule-1.
Biosensors & bioelectronics (2018-06-01)
Huiyun Yang, Haijun Wang, Chengyi Xiong, Yaqin Chai, Ruo Yuan
ABSTRACT
In this work, poly[9,9-dioctylfluorenyl-2,7-diyl] (PFO) dots is discovered to display an appealing dual enhancement effect for the electrochemiluminescence (ECL) system of N-(aminobutyl)-N-(ethylisoluminol)/hydrogen peroxide (ABEI/H2O2), which not only enhances the ECL intensity of ABEI but also catalyzes decomposition of H2O2 to further amplify the ECL signal of ABEI. Owing to the electronegative property of PFO dots, electropositive ABEI-PEI as ECL reagent could be adsorbed on their surface and thus form a novel luminescence emitter (ABEI-PEI-PFO dots) with high ECL efficiency based on electrostatic attraction. Meanwhile, the water solubility and stability of this emitter are improved in virtue of the amine-rich property of ECL reagent (ABEI-PEI), which could increase the luminous efficiency of ECL reaction in aqueous solution. To increase the electron transfer efficiency, Pt nanoparticles (PtNPs) supported on reduced graphene oxide nanosheets (RGOs) via a onepot synthetic strategy are chosen as immobilizing platform for the ECL emitter (ABEI-PEI-PFO dots). Herein, the obtained dual-amplifed ABEI-PEI-PFO dots-RGOs/PtNPs complex is served as an ideal nanocarrier to capture detection antibody (Ab2). According to sandwiched immunoreaction, a highly sensitive ECL immunosensor is constructed for the detection of kidney injury molecule-1 (KIM-1) with a linearity from 50 fg mL-1 to 1 ng mL-1 and a detection limit of 16.7 fg mL-1. The developed ECL emitter combining dual amplified property for signal enhancement purpose would provide new thought and potential for sensitive bioanalysis and clinical application.