Merck
  • 3D bioprinting of vascularized, heterogeneous cell-laden tissue constructs.
3D bioprinting of vascularized, heterogeneous cell-laden tissue constructs.
Advanced materials (Deerfield Beach, Fla.) (2014-02-20)
David B Kolesky, Ryan L Truby, A Sydney Gladman, Travis A Busbee, Kimberly A Homan, Jennifer A Lewis
ABSTRACT
A new bioprinting method is reported for fabricating 3D tissue constructs replete with vasculature, multiple types of cells, and extracellular matrix. These intricate, heterogeneous structures are created by precisely co-printing multiple materials, known as bioinks, in three dimensions. These 3D micro-engineered environments open new -avenues for drug screening and fundamental studies of wound healing, angiogenesis, and stem-cell niches.
Sigma-Aldrich
TissueFab® bioink kit, (Gel)ma Fibrin (UV/365), low endotoxin
Sigma-Aldrich
TissueFab® bioink kit, (Gel)ma Fibrin (Vis/405), low endotoxin