Merck
  • Acceleration of chronic wound healing by bio-inorganic polyphosphate: In vitro studies and first clinical applications.
Acceleration of chronic wound healing by bio-inorganic polyphosphate: In vitro studies and first clinical applications.
Theranostics (2022-01-07)
Hadrian Schepler, Meik Neufurth, Shunfeng Wang, Zhengding She, Heinz C Schröder, Xiaohong Wang, Werner E G Müller
ABSTRACT
The healing of chronic wounds is impaired by a lack of metabolic energy. In previous studies, we showed that physiological inorganic polyphosphate (polyP) is a generator of metabolic energy by forming ATP as a result of the enzymatic cleavage of the high-energy phosphoanhydride bonds of this polymer. Therefore, in the present study, we investigated whether the administration of polyP can substitute for the energy deficiency in chronic wound healing. Methods: PolyP was incorporated into collagen mats and applied in vitro and to patients in vivo. Results: (i) In vitro studies: Keratinocytes grown in vitro onto the polyP/collagen mats formed long microvilli to guide them to a favorable environment. HUVEC cells responded to polyP/collagen mats with an increased adhesion and migration propensity as well as penetration into the mats. (ii) In vivo - human clinical studies: In a "bench to bedside" process these promising in vitro results were translated from the laboratory into the clinic. In the proof-of-concept application, the engineered polyP/collagen mats were applied to chronic wounds in patients. Those mats impressively accelerated the re-epithelialization rate, with a reduction of the wound area to 65% after 3 weeks and to 36.6% and 22.5% after 6 and 9 weeks, respectively. Complete healing was achieved and no further treatment was necessary. Biopsy samples from the regenerating wound area showed predominantly myofibroblasts. The wound healing process was supported by the use of a polyP containing moisturizing solution. Conclusion: The results strongly recommend polyP as a beneficial component in mats for a substantial healing of chronic wounds.
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, suitable for hybridization, pH 7, ≥98%
Millipore
Microscope Slides, Accessories for filter holders for sample preparation.
Sigma-Aldrich
5-Bromo-4-chloro-3-indolyl phosphate disodium salt, ≥98% (HPLC)
Sigma-Aldrich
Anti-Cytokeratin AE1/AE3 Antibody, recognizes acidic & basic cytokeratins, clone AE1/AE3, clone AE1/AE3, Chemicon®, from mouse