Merck
  • PEG-PLGA copolymers: their structure and structure-influenced drug delivery applications.
PEG-PLGA copolymers: their structure and structure-influenced drug delivery applications.
Journal of controlled release : official journal of the Controlled Release Society (2014-03-29)
Keru Zhang, Xing Tang, Juan Zhang, Wei Lu, Xia Lin, Yu Zhang, Bin Tian, Hua Yang, Haibing He
ABSTRACT
In the paper, we begin by describing polyethylene glycol-poly lactic acid-co-glycolic acid (PEG-PLGA) which was chosen as a typical model copolymer for the construction of nano-sized drug delivery systems and also the types of PEG-PLGA copolymers that were eluted. Following this we examine the structure-influenced drug delivery applications including nanoparticles, micelles and hydrogels. After that, the preparation methods for nano-sized delivery systems are presented. In addition, the drug loading mode of PEG-PLGA micelles is divided into three aspects. Finally, the drug release profiles of PEG-PLGA micelles, both in terms of their in vitro and in vivo characteristics, are represented. PEG-PLGA copolymers are very suitable for the construction of micelles as carriers for insoluble drugs. This article reviews the structure and the different structure-influenced applications of PEG-PLGA copolymers, concentrating on the application of PEG-PLGA micelles.