Merck
  • Antagonists that demonstrate species differences in neurokinin-1 receptors.
Antagonists that demonstrate species differences in neurokinin-1 receptors.
Molecular pharmacology (1992-04-01)
K C Appell, B J Fragale, J Loscig, S Singh, B E Tomczuk
ABSTRACT
125I-Bolton-Hunter-substance P (125I-BH-SP) binding properties of three novel classes of neurokinin-1 (NK-1) receptor antagonists were investigated in tissues derived from humans, guinea pigs, and rats. 125I-BH-SP was shown to bind to a single class of binding sites, with similar dissociation constants, Kd, in human astrocytoma cells (U-373 MG), human urinary bladder, guinea pig forebrain, guinea pig ileum longitudinal smooth muscle, rat forebrain, and rat duodenum. In each tissue preparation, known peptide agonists and peptide antagonists yielded potencies typical for a NK-1 receptor profile, with little difference in binding properties between the various tissues. However, when the three classes of compounds, heterosteroids, cyanines, and modified peptides, were tested for their ability to displace 125I-BH-SP binding from the NK-1 receptor, very different binding profiles were observed. The heterosteroids were shown to be as much as 3 orders of magnitude more potent in tissues derived from rats than from humans or guinea pigs. A distinct species-dependent structure-activity relationship (SAR) was also observed for this class of compounds. Like the heterosteroids, the cyanines displaced 125I-BH-SP with 10-30-fold higher affinity in rat tissues than in human and guinea pig tissues. However, the SAR generated by the cyanines was comparable in all tissues studied. The modified peptides, on the other hand, were up to 10-100-fold more potent in human and guinea pig than rat tissues, producing a SAR that differed between the various species. No differences in binding properties between central nervous system and peripheral tissues from the same species were seen with these compounds. These results provide evidence for species differences in NK-1 receptors in humans, guinea pigs, and rats. Because it is known that there exists great sequence identity between rat and human NK-1 receptors, it is hypothesized that key amino acid changes or different lipid environments within the transmembrane binding region of the receptor may account for the observed species difference. Furthermore, this study emphasizes that caution is necessary in the choice of species to be used in development programs targeted towards therapeutic entities in the NK-1 receptor antagonist area.
Sigma-Aldrich
WIN 62,577, solid