Merck
  • Telomerase reverse transcriptase activates transcription of miR500A to inhibit Hedgehog signalling and promote cell invasiveness.
Telomerase reverse transcriptase activates transcription of miR500A to inhibit Hedgehog signalling and promote cell invasiveness.
Molecular oncology (2021-03-14)
Manuel Bernabé-García, Elena Martínez-Balsalobre, Diana García-Moreno, Jesús García-Castillo, Beatriz Revilla-Nuin, Elena Blanco-Alcaina, Victoriano Mulero, Francisca Alcaraz-Pérez, María L Cayuela
ABSTRACT
Telomerase reverse transcriptase (TERT) maintains telomere homeostasis, thus ensuring chromosome stability and cell proliferation. In addition, several telomere-independent functions of human TERT have been described. In this study, we report that TERT binds directly to the TCF binding elements located upstream of the oncomiR miR500A, and induces its transcription. This function was independent of the telomerase activity, as shown with experiments using catalytically inactive TERT and inhibitors of TERT and the TERT RNA component. miR500A was in turn found to target three key components of the Hedgehog signalling pathway: Patched 1; Gli family zinc finger 3; and Cullin 3, thereby promoting cell invasion. Our results point to the crucial role of the TERT-miR500A-Hedgehog axis in tumour aggressiveness and highlight the therapeutic potential of targeting noncanonical TERT functions in cancer.
Sigma-Aldrich
N-(2-Mercaptopropionyl)glycine
Sigma-Aldrich
TRAPeze1X CHAPS Lysis Buffer, 1X CHAPS Lysis Buffer is a component of the gel-and ELISA-based TRAPeze Telomerase Detection Kits & is used in the extraction step of the procedure.
Supelco
Ethyl 3-aminobenzoate methanesulfonate salt, analytical standard
Sigma-Aldrich
Paraformaldehyde, reagent grade, crystalline
Sigma-Aldrich
Protease Inhibitor Cocktail powder, for general use, lyophilized powder