Merck
  • Estrogen receptors orchestrate cell growth and differentiation to facilitate liver regeneration.
Estrogen receptors orchestrate cell growth and differentiation to facilitate liver regeneration.
Theranostics (2018-05-19)
Ta-Lun Kao, Yu-Ping Kuan, Wei-Chung Cheng, Wei-Chun Chang, Long-Bin Jeng, Shuyuan Yeh, Wen-Lung Ma
ABSTRACT
Background and Aims: Improving liver regeneration (LR) capacity and thereby liver function reserve is a critical bridging strategy for managing liver failure patients. Since estrogen signaling may participate in LR, our aim was to characterize the roles of ERα and ERβ in LR. Methods: LR capacity and estradiol levels following 2/3rd partial hepatectomy (PHx) were compared in ERα-KO or ERβ-KO vs. wildtype mice. The ERα- or ERβ-related transcriptome and interactome were analyzed from regenerating livers, and then bioinformatics was used for pathway discovery and analysis of interactome-transcriptome relationships. Human hepatic progenitors (HepRG cells) and mouse Hepa1-6 hepatocytes were used to elucidate molecular interactions and functions. Results: This paper demonstrated that estrogen signals orchestrate hepatic repopulation and differentiation via distinct transcriptome patterns governed by ERα or ERβ. Cell repopulation pathway was associated with the ERα-transcriptome, but cell differentiation and metabolic function were associated with the ERβ transcriptome. Mechanistic studies linking ERs interactomes and transcriptomes discovered that ERα-Chd1 interaction promoted cell growth by upregulating Ssxb6, Crygc, and Cst1; and, ERβ-Ube3a interaction facilitated hepatic progenitor cell differentiation to hepatocytes and cholangiocytes, specifically by upregulating Ifna5. Conclusions: ERα and ERβ orchestrate liver cell proliferation and differentiation respectively, thereby promoting LR.
Sigma-Aldrich
1,4-Dithioerythritol, BioReagent, for molecular biology, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ammonium bicarbonate, ReagentPlus®, ≥99.0%