Merck
  • A dataset for the analysis of antibody response to glycan alpha-Gal in individuals with immune-mediated disorders.
A dataset for the analysis of antibody response to glycan alpha-Gal in individuals with immune-mediated disorders.
F1000Research (2021-08-21)
José de la Fuente, José Miguel Urra, Marinela Contreras, Iván Pacheco, Elisa Ferreras-Colino, Ernesto Doncel-Pérez, Isabel G Fernández de Mera, Margarita Villar, Carmen M Cabrera, Cesar Gómez Hernando, Eduardo Vargas Baquero, Javier Blanco García, Javier Rodríguez Gómez, Alberto Velayos Galán, Francisco Feo Brito, Elisa Gómez Torrijos, Alejandro Cabezas-Cruz, Christian Gortázar
ABSTRACT
Humans evolved by losing the capacity to synthesize the glycan Galα1-3Galβ1-(3)4GlcNAc-R (α-Gal), which resulted in the development of a protective response mediated by anti-α-Gal IgM/IgG/IgA antibodies against pathogens containing this modification on membrane proteins. As an evolutionary trade-off, humans can develop the alpha-Gal syndrome (AGS), a recently diagnosed disease mediated by anti-α-Gal IgE antibodies and associated with allergic reactions to mammalian meat consumption and tick bites. However, the anti-α-Gal antibody response may be associated with other immune-mediated disorders such as those occurring in patients with COVID-19 and Guillain-Barré syndrome (GBS). Here, we provide a dataset (209 entries) on the IgE/IgM/IgG/IgA anti-α-Gal antibody response in healthy individuals and patients diagnosed with AGS, tick-borne allergies, GBS and COVID-19. The data allows correlative analyses of the anti-α-Gal antibody response with factors such as patient and clinical characteristics, record of tick bites, blood group, age and sex. These analyses could provide insights into the role of anti-α-Gal antibody response in disease symptomatology and possible protective mechanisms.
Sigma-Aldrich
Anti-Human IgM (μ-chain specific) antibody produced in goat, whole antiserum
Sigma-Aldrich
Anti-Human IgE (ε-chain specific) antibody produced in goat, affinity isolated antibody, lyophilized powder
Sigma-Aldrich
Anti-Human IgG (Fc specific) antibody produced in goat, affinity isolated antibody, buffered aqueous solution