Merck
  • Secreted gelsolin inhibits DNGR-1-dependent cross-presentation and cancer immunity.
Secreted gelsolin inhibits DNGR-1-dependent cross-presentation and cancer immunity.
Cell (2021-06-04)
Evangelos Giampazolias, Oliver Schulz, Kok Haw Jonathan Lim, Neil C Rogers, Probir Chakravarty, Naren Srinivasan, Oliver Gordon, Ana Cardoso, Michael D Buck, Enzo Z Poirier, Johnathan Canton, Santiago Zelenay, Stefano Sammicheli, Natalia Moncaut, Sunita Varsani-Brown, Ian Rosewell, Caetano Reis E Sousa
ABSTRACT
Cross-presentation of antigens from dead tumor cells by type 1 conventional dendritic cells (cDC1s) is thought to underlie priming of anti-cancer CD8+ T cells. cDC1 express high levels of DNGR-1 (a.k.a. CLEC9A), a receptor that binds to F-actin exposed by dead cell debris and promotes cross-presentation of associated antigens. Here, we show that secreted gelsolin (sGSN), an extracellular protein, decreases DNGR-1 binding to F-actin and cross-presentation of dead cell-associated antigens by cDC1s. Mice deficient in sGsn display increased DNGR-1-dependent resistance to transplantable tumors, especially ones expressing neoantigens associated with the actin cytoskeleton, and exhibit greater responsiveness to cancer immunotherapy. In human cancers, lower levels of intratumoral sGSN transcripts, as well as presence of mutations in proteins associated with the actin cytoskeleton, are associated with signatures of anti-cancer immunity and increased patient survival. Our results reveal a natural barrier to cross-presentation of cancer antigens that dampens anti-tumor CD8+ T cell responses.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2-Peroxidase (HRP) antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
ExtrAvidin® –Alkaline Phosphatase, buffered aqueous solution
Sigma-Aldrich
Anti-β-Actin−Peroxidase antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Roche
DNase I, from bovine pancreas
Sigma-Aldrich
Monoclonal Anti-Actin antibody produced in mouse, clone AC-40, ascites fluid
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥98% (agarose gel electrophoresis)
Roche
CPRG, Chlorophenol red-β-D-galactopyranoside