Merck
  • A novel inactivated whole-cell Pseudomonas aeruginosa vaccine that acts through the cGAS-STING pathway.
A novel inactivated whole-cell Pseudomonas aeruginosa vaccine that acts through the cGAS-STING pathway.
Signal transduction and targeted therapy (2021-10-02)
Cuicui Ma, Xiao Ma, Boguang Jiang, Hailong Pan, Xueyuan Liao, Li Zhang, Wenfang Li, Yingjie Luo, Zhixue Shen, Xingjun Cheng, Mao Lian, Zhenling Wang
ABSTRACT
Pseudomonas aeruginosa infection continues to be a major threat to global public health, and new safe and efficacious vaccines are needed for prevention of infections caused by P. aeruginosa. X-ray irradiation has been used to prepare whole-cell inactivated vaccines against P. aeruginosa infection. However, the immunological mechanisms of X-ray-inactivated vaccines are still unclear and require further investigation. Our previous study found that an X-ray-inactivated whole-cell vaccine could provide protection against P. aeruginosa by boosting T cells. The aim of the present study was to further explore the immunological mechanisms of the vaccine. Herein, P. aeruginosa PAO1, a widely used laboratory strain, was utilized to prepare the vaccine, and we found nucleic acids and 8-hydroxyguanosine in the supernatant of X-ray-inactivated PAO1 (XPa). By detecting CD86, CD80, and MHCII expression, we found that XPa fostered dentritic cell (DC) maturation by detecting. XPa stimulated the cGAS-STING pathway as well as Toll-like receptors in DCs in vitro, and DC finally underwent apoptosis and pyroptosis after XPa stimulation. In addition, DC stimulated by XPa induced CD8+ T-cell proliferation in vitro and generated immunologic memory in vivo. Moreover, XPa vaccination induced both Th1 and Th2 cytokine responses in mice and reduced the level of inflammatory factors during infection. XPa protected mice in pneumonia models from infection with PAO1 or multidrug-resistant clinical isolate W9. Chronic obstructive pulmonary disease (COPD) mice immunized with XPa could resist PAO1 infection. Therefore, a new mechanism of an X-ray-inactivated whole-cell vaccine against P. aeruginosa infection was discovered in this study.
Sigma-Aldrich
Lipopolysaccharides from Pseudomonas aeruginosa 10, purified by phenol extraction
Sigma-Aldrich
Elastase from porcine pancreas, Type I, ≥4.0 units/mg protein
Millipore
MILLIPLEX® TGFß Magnetic Bead 3 Plex Kit - Immunology Multiplex Assay, for quantification of TGFß1, TGFß2 and TGFß3 in multiple species
Sigma-Aldrich
2′,3′-cGAMP sodium salt, ≥98% (HPLC)