Merck
  • Transcriptional repression by FEZF2 restricts alternative identities of cortical projection neurons.
Transcriptional repression by FEZF2 restricts alternative identities of cortical projection neurons.
Cell reports (2021-06-24)
Jeremiah Tsyporin, David Tastad, Xiaokuang Ma, Antoine Nehme, Thomas Finn, Liora Huebner, Guoping Liu, Daisy Gallardo, Amr Makhamreh, Jacqueline M Roberts, Solomon Katzman, Nenad Sestan, Susan K McConnell, Zhengang Yang, Shenfeng Qiu, Bin Chen
ABSTRACT
Projection neuron subtype identities in the cerebral cortex are established by expressing pan-cortical and subtype-specific effector genes that execute terminal differentiation programs bestowing neurons with a glutamatergic neuron phenotype and subtype-specific morphology, physiology, and axonal projections. Whether pan-cortical glutamatergic and subtype-specific characteristics are regulated by the same genes or controlled by distinct programs remains largely unknown. Here, we show that FEZF2 functions as a transcriptional repressor, and it regulates subtype-specific identities of both corticothalamic and subcerebral neurons by selectively repressing expression of genes inappropriate for each neuronal subtype. We report that TLE4, specifically expressed in layer 6 corticothalamic neurons, is recruited by FEZF2 to inhibit layer 5 subcerebral neuronal genes. Together with previous studies, our results indicate that a cortical glutamatergic identity is specified by multiple parallel pathways active in progenitor cells, whereas projection neuron subtype-specific identity is achieved through selectively repressing genes associated with alternate identities in differentiating neurons.
Sigma-Aldrich
Anti-FOSL2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Copper(II) sulfate, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
(+)-Sodium L-ascorbate, crystalline, ≥98%
Millipore
Anti-FLAG® M2 Magnetic Beads, affinity isolated antibody
Roche
DIG RNA Labeling Mix, sufficient for 20 reactions, solution
Roche
NBT/BCIP Stock Solution
Sigma-Aldrich
GenElute HP Plasmid Miniprep Kit, sufficient for 350 preparations