Merck
  • Single-cell ATAC and RNA sequencing reveal pre-existing and persistent cells associated with prostate cancer relapse.
Single-cell ATAC and RNA sequencing reveal pre-existing and persistent cells associated with prostate cancer relapse.
Nature communications (2021-09-08)
S Taavitsainen, N Engedal, S Cao, F Handle, A Erickson, S Prekovic, D Wetterskog, T Tolonen, E M Vuorinen, A Kiviaho, R Nätkin, T Häkkinen, W Devlies, S Henttinen, R Kaarijärvi, M Lahnalampi, H Kaljunen, K Nowakowska, H Syvälä, M Bläuer, P Cremaschi, F Claessens, T Visakorpi, T L J Tammela, T Murtola, K J Granberg, A D Lamb, K Ketola, I G Mills, G Attard, W Wang, M Nykter, A Urbanucci
ABSTRACT
Prostate cancer is heterogeneous and patients would benefit from methods that stratify those who are likely to respond to systemic therapy. Here, we employ single-cell assays for transposase-accessible chromatin (ATAC) and RNA sequencing in models of early treatment response and resistance to enzalutamide. In doing so, we identify pre-existing and treatment-persistent cell subpopulations that possess regenerative potential when subjected to treatment. We find distinct chromatin landscapes associated with enzalutamide treatment and resistance that are linked to alternative transcriptional programs. Transcriptional profiles characteristic of persistent cells are able to stratify the treatment response of patients. Ultimately, we show that defining changes in chromatin and gene expression in single-cell populations from pre-clinical models can reveal as yet unrecognized molecular predictors of treatment response. This suggests that the application of single-cell methods with high analytical resolution in pre-clinical models may powerfully inform clinical decision-making.
Sigma-Aldrich
Ala-Gln, 200 mM, solution, sterile-filtered, Biotechnology Performance Certified
Sigma-Aldrich
D-(+)-Glucose solution, 45% in H2O, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Trypsin-EDTA solution, 1 ×, sterile; sterile-filtered, BioReagent, suitable for cell culture, 0.5 g porcine trypsin and 0.2 g EDTA, 4Na per liter of Hanks′ Balanced Salt Solution with phenol red
Sigma-Aldrich
RPMI-1640 Medium, With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture