Merck
  • Nanoluciferase complementation-based bioreporter reveals the importance of N-linked glycosylation of SARS-CoV-2 S for viral entry.
Nanoluciferase complementation-based bioreporter reveals the importance of N-linked glycosylation of SARS-CoV-2 S for viral entry.
Molecular therapy : the journal of the American Society of Gene Therapy (2021-02-13)
Taha Azad, Ragunath Singaravelu, Zaid Taha, Taylor R Jamieson, Stephen Boulton, Mathieu J F Crupi, Nikolas T Martin, Emily E F Brown, Joanna Poutou, Mina Ghahremani, Adrian Pelin, Kazem Nouri, Reza Rezaei, Christopher Boyd Marshall, Masahiro Enomoto, Rozanne Arulanandam, Nouf Alluqmani, Reuben Samson, Anne-Claude Gingras, D William Cameron, Peter A Greer, Carolina S Ilkow, Jean-Simon Diallo, John C Bell
ABSTRACT
The ongoing COVID-19 pandemic has highlighted the immediate need for the development of antiviral therapeutics targeting different stages of the SARS-CoV-2 life cycle. We developed a bioluminescence-based bioreporter to interrogate the interaction between the SARS-CoV-2 viral spike (S) protein and its host entry receptor, angiotensin-converting enzyme 2 (ACE2). The bioreporter assay is based on a nanoluciferase complementation reporter, composed of two subunits, large BiT and small BiT, fused to the S receptor-binding domain (RBD) of the SARS-CoV-2 S protein and ACE2 ectodomain, respectively. Using this bioreporter, we uncovered critical host and viral determinants of the interaction, including a role for glycosylation of asparagine residues within the RBD in mediating successful viral entry. We also demonstrate the importance of N-linked glycosylation to the RBD's antigenicity and immunogenicity. Our study demonstrates the versatility of our bioreporter in mapping key residues mediating viral entry as well as screening inhibitors of the ACE2-RBD interaction. Our findings point toward targeting RBD glycosylation for therapeutic and vaccine strategies against SARS-CoV-2.
Sigma-Aldrich
Lectin from Galanthus nivalis (snowdrop), lyophilized powder
Sigma-Aldrich
Lectin from Lens culinaris (lentil), Isoelectric focusing marker, pI (1) 8.2, (2) 8.6, (3) 8.8
Sigma-Aldrich
Anti-HA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Concanavalin A from Canavalia ensiformis (Jack bean), Type VI, lyophilized powder
Sigma-Aldrich
Lectin from Pisum sativum (pea), lyophilized powder
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)