Merck
  • Evaluation of the PEΔIII-LC3-KDEL3 Chimeric Protein of Entamoeba histolytica-Lectin as a Vaccine Candidate against Amebic Liver Abscess.
Evaluation of the PEΔIII-LC3-KDEL3 Chimeric Protein of Entamoeba histolytica-Lectin as a Vaccine Candidate against Amebic Liver Abscess.
Journal of immunology research (2021-04-08)
Sandra L Martínez-Hernández, Viridiana M Becerra-González, Martín H Muñoz-Ortega, Víctor M Loera-Muro, Manuel E Ávila-Blanco, Marina N Medina-Rosales, Javier Ventura-Juárez
ABSTRACT
Entamoeba histolytica is an intestinal parasite that causes dysentery and amebic liver abscess. E. histolytica has the capability to invade host tissue by union of virulence factor Gal/GalNAc lectin; this molecule induces an adherence-inhibitory antibody response as well as to protect against amebic liver abscess (ALA). The present work showed the effect of the immunization with PEΔIII-LC3-KDEL3 recombinant protein. In vitro, this candidate vaccine inhibited adherence of E. histolytica trophozoites to HepG2 cell monolayer, avoiding the cytolysis, and in a hamster model, we observed a vaccine-induced protection against the damage to tissue liver and the inhibition of uncontrolled inflammation. PEΔIII-LC3-KDEL3 reduced the expression of TNF-α, IL-1β, and NF-κB in all immunized groups at 4- and 7-day postinfection. The levels of IL-10, FOXP3, and IFN-γ were elevated at 7 days. The immunohistochemistry assay confirmed this result, revealing an elevated quantity of +IFN-γ cells in the liver tissue. ALA formation in hamsters immunized was minimal, and few trophozoites were identified. Hence, immunization with PEΔIII-LC3-KDEL3 herein prevented invasive amebiasis, avoided an acute proinflammatory response, and activated a protective response within a short time. Finally, this recombinant protein induced an increase of serum IgG.
Sigma-Aldrich
Anti-Hamster IgG (whole molecule) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution