Merck
  • GAGE mediates radio resistance in cervical cancers via the regulation of chromatin accessibility.
GAGE mediates radio resistance in cervical cancers via the regulation of chromatin accessibility.
Cell reports (2021-09-02)
Dawn Sijin Nin, Caryn Wujanto, Tuan Zea Tan, Diana Lim, J Mirjam A Damen, Kuan-Yi Wu, Ziyu Melvin Dai, Zheng-Wei Lee, Shabana Binte Idres, Yiat Horng Leong, Sudhakar Jha, Joseph Soon-Yau Ng, Jeffrey J H Low, Shih-Chung Chang, David Shao Peng Tan, Wei Wu, Bok Ai Choo, Lih-Wen Deng
ABSTRACT
Radiotherapy (RT) resistance is a major cause of treatment failure in cancers that use definitive RT as their primary treatment modality. This study identifies the cancer/testis (CT) antigen G antigen (GAGE) as a mediator of radio resistance in cervical cancers. Elevated GAGE expression positively associates with de novo RT resistance in clinical samples. GAGE, specifically the GAGE12 protein variant, confers RT resistance through synemin-dependent chromatin localization, promoting the association of histone deacetylase 1/2 (HDAC1/2) to its inhibitor actin. This cumulates to elevated histone 3 lysine 56 acetylation (H3K56Ac) levels, increased chromatin accessibility, and improved DNA repair efficiency. Molecular or pharmacological disruption of the GAGE-associated complex restores radiosensitivity. Molecularly, this study demonstrates the role of GAGE in the regulation of chromatin dynamics. Clinically, this study puts forward the utility of GAGE as a pre-screening biomarker to identify poor responders at initial diagnosis and the therapeutic potential of agents that target GAGE and its associated complex in combination with radiotherapy to improve outcomes.
Sigma-Aldrich
Anti-HDAC8 Antibody, serum, Upstate®
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Anti-p300 CT Antibody, clone RW128, clone RW128, Upstate®, from mouse
Sigma-Aldrich
Anti-PP2A Antibody, C subunit, clone 1D6, clone 1D6, Upstate®, from mouse