Merck
  • Improving cell-free glycoprotein synthesis by characterizing and enriching native membrane vesicles.
Improving cell-free glycoprotein synthesis by characterizing and enriching native membrane vesicles.
Nature communications (2021-04-24)
Jasmine M Hershewe, Katherine F Warfel, Shaelyn M Iyer, Justin A Peruzzi, Claretta J Sullivan, Eric W Roth, Matthew P DeLisa, Neha P Kamat, Michael C Jewett
ABSTRACT
Cell-free gene expression (CFE) systems from crude cellular extracts have attracted much attention for biomanufacturing and synthetic biology. However, activating membrane-dependent functionality of cell-derived vesicles in bacterial CFE systems has been limited. Here, we address this limitation by characterizing native membrane vesicles in Escherichia coli-based CFE extracts and describing methods to enrich vesicles with heterologous, membrane-bound machinery. As a model, we focus on bacterial glycoengineering. We first use multiple, orthogonal techniques to characterize vesicles and show how extract processing methods can be used to increase concentrations of membrane vesicles in CFE systems. Then, we show that extracts enriched in vesicle number also display enhanced concentrations of heterologous membrane protein cargo. Finally, we apply our methods to enrich membrane-bound oligosaccharyltransferases and lipid-linked oligosaccharides for improving cell-free N-linked and O-linked glycoprotein synthesis. We anticipate that these methods will facilitate on-demand glycoprotein production and enable new CFE systems with membrane-associated activities.
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Putrescine dihydrochloride, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Guanosine 5′-triphosphate sodium salt hydrate, ≥95% (HPLC), powder
Sigma-Aldrich
Coenzyme A sodium salt hydrate, cofactor for acyl transfer
Sigma-Aldrich
L-Glutamic acid hemimagnesium salt tetrahydrate, ≥98.0% (NT)
Roche
Phosphoenol-pyruvate, 97% (PEP-K), pkg of 1 g
Sigma-Aldrich
Uridine 5′-triphosphate trisodium salt hydrate, from yeast, Type III, ≥96%
Sigma-Aldrich
L-Glutamic acid potassium salt monohydrate, ≥99% (HPLC), powder
Sigma-Aldrich
Cytidine 5′-triphosphate disodium salt, ≥95%
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt hydrate, Grade I, ≥99%, from microbial
Sigma-Aldrich
Spermidine, ≥99% (GC)