Merck
  • An In Vitro Batch-culture Model to Estimate the Effects of Interventional Regimens on Human Fecal Microbiota.
An In Vitro Batch-culture Model to Estimate the Effects of Interventional Regimens on Human Fecal Microbiota.
Journal of visualized experiments : JoVE (2019-08-20)
Shokouh Ahmadi, Shaohua Wang, Ravinder Nagpal, Rabina Mainali, Sabihe Soleimanian-Zad, Dalane Kitzman, Hariom Yadav
ABSTRACT
The emerging role of the gut microbiome in several human diseases demands a breakthrough of new tools, techniques and technologies. Such improvements are needed to decipher the utilization of microbiome modulators for human health benefits. However, the large-scale screening and optimization of modulators to validate microbiome modulation and predict related health benefits may be practically difficult due to the need for large number of animals and/or human subjects. To this end, in vitro or ex vivo models can facilitate preliminary screening of microbiome modulators. Herein, it is optimized and demonstrated an ex vivo fecal microbiota culture system that can be used for examining the effects of various interventions of gut microbiome modulators including probiotics, prebiotics and other food ingredients, aside from nutraceuticals and drugs, on the diversity and composition of the human gut microbiota. Inulin, one of the most widely studied prebiotic compounds and microbiome modulators, is used as an example here to examine its effect on the healthy fecal microbiota composition and its metabolic activities, such as fecal pH and the fecal levels of organic acids including lactate and short-chain fatty acids (SCFAs). The protocol may be useful for studies aimed at estimating the effects of different interventions of modulators on fecal microbiota profiles and at predicting their health impacts.
Sigma-Aldrich
L-Cysteine hydrochloride hydrate, 99%
Sigma-Aldrich
Vitamin B12, ≥98%
Sigma-Aldrich
Resazurin sodium salt, powder, BioReagent
Sigma-Aldrich
Hemin, from bovine, ≥90%
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
Calcium chloride dihydrate, for molecular biology, ≥99.0%
Sigma-Aldrich
Zinc sulfate heptahydrate, BioReagent, suitable for cell culture
Sigma-Aldrich
Nicotinic acid, ≥98%
Sigma-Aldrich
Biotin, ≥99% (HPLC), lyophilized powder
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Valeric acid, ≥99%