Merck
  • Step-by-step assembly and testing of a low-cost bioprinting solution for research and educational purposes.
Step-by-step assembly and testing of a low-cost bioprinting solution for research and educational purposes.
MethodsX (2020-12-31)
John Robert Honiball, Michael Sean Pepper, Earl Prinsloo
ABSTRACT
Bioprinting is a rapidly expanding technology with the ability to fabricate in vitro three-dimensional (3D) tissues in a layer-by-layer manner to ultimately produce a living tissue which physiologically resembles native in vivo tissue functionality. Unfortunately, large costs associated with commercially available bioprinters severely limit access to the technology. We investigated the potential for modifying a low-cost commercially available RepRap Prusa iteration 3 (i3) 3D printer with an open-source syringe-housed microextrusion print-head unit (universal paste extruder by Richard Horne, RichRap), that allowed for controlled deposition of cell-laden bioinks and Freeform Reversible Embedding of Suspended Hydrogels (FRESH) method-based printing.
Sigma-Aldrich
Antibiotic Antimycotic Solution (100×), Stabilized, with 10,000 units penicillin, 10 mg streptomycin and 25 μg amphotericin B per mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Minimum Essential Medium Eagle, Modified, with Earle′s salts and reduced NaHCO3. without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sigma-Aldrich
Rosiglitazone, ≥98% (HPLC)
Sigma-Aldrich
Insulin human, recombinant, expressed in yeast, γ-irradiated, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Phosphate Buffered Saline, Modified, without calcium chloride and magnesium chloride, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
L-Glutamine solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture