Merck
  • Stress-primed secretory autophagy promotes extracellular BDNF maturation by enhancing MMP9 secretion.
Stress-primed secretory autophagy promotes extracellular BDNF maturation by enhancing MMP9 secretion.
Nature communications (2021-08-01)
Silvia Martinelli, Elmira A Anderzhanova, Thomas Bajaj, Svenja Wiechmann, Frederik Dethloff, Katja Weckmann, Daniel E Heinz, Tim Ebert, Jakob Hartmann, Thomas M Geiger, Michael Döngi, Kathrin Hafner, Max L Pöhlmann, Lee Jollans, Alexandra Philipsen, Susanne V Schmidt, Ulrike Schmidt, Giuseppina Maccarrone, Valentin Stein, Felix Hausch, Christoph W Turck, Mathias V Schmidt, Anne-Kathrin Gellner, Bernhard Kuster, Nils C Gassen
ABSTRACT
The stress response is an essential mechanism for maintaining homeostasis, and its disruption is implicated in several psychiatric disorders. On the cellular level, stress activates, among other mechanisms, autophagy that regulates homeostasis through protein degradation and recycling. Secretory autophagy is a recently described pathway in which autophagosomes fuse with the plasma membrane rather than with lysosomes. Here, we demonstrate that glucocorticoid-mediated stress enhances secretory autophagy via the stress-responsive co-chaperone FK506-binding protein 51. We identify the matrix metalloproteinase 9 (MMP9) as one of the proteins secreted in response to stress. Using cellular assays and in vivo microdialysis, we further find that stress-enhanced MMP9 secretion increases the cleavage of pro-brain-derived neurotrophic factor (proBDNF) to its mature form (mBDNF). BDNF is essential for adult synaptic plasticity and its pathway is associated with major depression and posttraumatic stress disorder. These findings unravel a cellular stress adaptation mechanism that bears the potential of opening avenues for the understanding of the pathophysiology of stress-related disorders.
Sigma-Aldrich
Anti-SNAP23 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
TWEEN® 20, viscous liquid, suitable for cell culture
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Protease Inhibitor Cocktail powder, for general use, lyophilized powder
Sigma-Aldrich
SIGMAFAST Protease Inhibitor Cocktail Tablets, EDTA-Free, for use in purification of Histidine-tagged proteins
Sigma-Aldrich
Anti-GAPDH Mouse mAb (6C5), liquid, clone 6C5, Calbiochem®
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Millipore
FLAG® Peptide, lyophilized powder