Merck
  • Direct Peptide Cyclization and One-Pot Modification Using the MeDbz Linker.
Direct Peptide Cyclization and One-Pot Modification Using the MeDbz Linker.
The Journal of organic chemistry (2018-08-07)
Bengt H Gless, Christian A Olsen
ABSTRACT
The one-pot synthesis and modification of cyclic peptides through a self-cleaving on-resin protocol is described. We apply Dawson's MeDbz linker to achieve direct intramolecular peptide cyclization by thioesterification followed by S → N acyl shift. This native chemical ligation approach requires no activating additive and allows direct modification of the crude cyclic peptides in one-pot. The strategy was applied to synthesize 5 cyclic peptide natural products of varying ring size. Finally, one-pot modifications include desulfurization, fluorophore conjugation, and intramolecular disulfide formation.
Sigma-Aldrich
SuTEx1-alkyne, ≥95%
Sigma-Aldrich
OxMet2-alkyne
Sigma-Aldrich
HMP-alkyne, ≥95%
Sigma-Aldrich
CP-alkyne, ≥95%