Merck
Avian mud nest architecture by self-secreted saliva.
Proceedings of the National Academy of Sciences of the United States of America (2021-01-13)
Yeonsu Jung, Sohyun Jung, Sang-Im Lee, Wonjung Kim, Ho-Young Kim
ABSTRACT
Mud nests built by swallows (Hirundinidae) and phoebes (Sayornis) are stable granular piles attached to cliffs, walls, or ceilings. Although these birds have been observed to mix saliva with incohesive mud granules, how such biopolymer solutions provide the nest with sufficient strength to support the weight of the residents as well as its own remains elusive. Here, we elucidate the mechanism of strong granular cohesion by the viscoelastic paste of bird saliva through a combination of theoretical analysis and experimental measurements in both natural and artificial nests. Our mathematical model considering the mechanics of mud nest construction allows us to explain the biological observation that all mud-nesting bird species should be lightweight.
Sigma-Aldrich
Mucin from porcine stomach, Type II
Sigma-Aldrich
Polyethylene glycol solution, BioUltra, for molecular biology, 1,000, ~50% in H2O
Sigma-Aldrich
Kaolinite, natural
Sigma-Aldrich
Poly(ethylene oxide), average Mv 100,000 (nominal), powder