Merck
  • Metastatic colorectal cancer cells maintain the TGFβ program and use TGFBI to fuel angiogenesis.
Metastatic colorectal cancer cells maintain the TGFβ program and use TGFBI to fuel angiogenesis.
Theranostics (2021-01-08)
Barbara Chiavarina, Brunella Costanza, Roberto Ronca, Arnaud Blomme, Sara Rezzola, Paola Chiodelli, Ambre Giguelay, Guillame Belthier, Gilles Doumont, Gaetan Van Simaeys, Simon Lacroix, Takehiko Yokobori, Bilguun Erkhem-Ochir, Patrick Balaguer, Vincent Cavailles, Eric Fabbrizio, Emmanuel Di Valentin, Stephanie Gofflot, Olivier Detry, Guy Jerusalem, Serge Goldman, Philippe Delvenne, Akeila Bellahcène, Julie Pannequin, Vincent Castronovo, Andrei Turtoi
ABSTRACT
Colorectal cancer (CRC) cells are traditionally considered unresponsive to TGFβ due to mutations in the receptors and/or downstream signaling molecules. TGFβ influences CRC cells only indirectly via stromal cells, such as cancer-associated fibroblasts. However, CRC cell ability to directly respond to TGFβ currently remains unexplored. This represents a missed opportunity for diagnostic and therapeutic interventions. Methods: We examined whether cancer cells from primary CRC and liver metastases respond to TGFβ by inducing TGFβ-induced protein ig-h3 (TGFBI) expression, and the contribution of canonical and non-canonical TGFβ signaling pathways to this effect. We then investigated in vitro and in vivo TGFBI impact on metastasis formation and angiogenesis. Using patient serum samples and an orthotopic mouse model of CRC liver metastases we assessed the diagnostic/tumor targeting value of novel antibodies against TGFBI. Results: Metastatic CRC cells, such as circulating tumor cells, directly respond to TGFβ. These cells were characterized by the absence of TGFβ receptor mutations and the frequent presence of p53 mutations. The pro-tumorigenic program orchestrated by TGFβ in CRC cells was mediated through TGFBI, the expression of which was positively regulated by non-canonical TGFβ signaling cascades. TGFBI inhibition was sufficient to significantly reduce liver metastasis formation in vivo. Moreover, TGFBI pro-tumorigenic function was linked to its ability to stimulate angiogenesis. TGFBI levels were higher in serum samples from untreated patients with CRC than in patients who were receiving chemotherapy. A radiolabeled anti-TGFBI antibody selectively targeted metastatic lesions in vivo, underscoring its diagnostic and therapeutic potential. Conclusions: TGFβ signaling in CRC cells directly contributes to their metastatic potential and stromal cell-independence. Proteins downstream of activated TGFβ, such as TGFBI, represent novel diagnostic and therapeutic targets for more specific anti-metastatic therapies.
Sigma-Aldrich
Mayer′s Hematoxylin Solution
Sigma-Aldrich
SB 202190, ≥98% (HPLC)
Sigma-Aldrich
MISSION® pLKO.1-puro eGFP shRNA Control Plasmid DNA, shRNA sequence targeting eGFP
Sigma-Aldrich
SP600125, ≥98% (HPLC)
Sigma-Aldrich
Bay 11-7082, ≥98% (HPLC), powder
Sigma-Aldrich
PD 98059
Roche
Transforming Growth Factor-β1, human (hTGF-β1), recombinant (CHO cells)