Merck
  • CENP-A overexpression promotes aneuploidy with karyotypic heterogeneity.
CENP-A overexpression promotes aneuploidy with karyotypic heterogeneity.
The Journal of cell biology (2021-02-24)
Roshan L Shrestha, Austin Rossi, Darawalee Wangsa, Ann K Hogan, Kimberly S Zaldana, Evelyn Suva, Yang Jo Chung, Chelsea L Sanders, Simone Difilippantonio, Tatiana S Karpova, Baktiar Karim, Daniel R Foltz, Daniele Fachinetti, Peter D Aplan, Thomas Ried, Munira A Basrai
ABSTRACT
Chromosomal instability (CIN) is a hallmark of many cancers. Restricting the localization of centromeric histone H3 variant CENP-A to centromeres prevents CIN. CENP-A overexpression (OE) and mislocalization have been observed in cancers and correlate with poor prognosis; however, the molecular consequences of CENP-A OE on CIN and aneuploidy have not been defined. Here, we show that CENP-A OE leads to its mislocalization and CIN with lagging chromosomes and micronuclei in pseudodiploid DLD1 cells and xenograft mouse model. CIN is due to reduced localization of proteins to the kinetochore, resulting in defects in kinetochore integrity and unstable kinetochore-microtubule attachments. CENP-A OE contributes to reduced expression of cell adhesion genes and higher invasion of DLD1 cells. We show that CENP-A OE contributes to aneuploidy with karyotypic heterogeneity in human cells and xenograft mouse model. In summary, our results provide a molecular link between CENP-A OE and aneuploidy, and suggest that karyotypic heterogeneity may contribute to the aggressive phenotype of CENP-A-overexpressing cancers.
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
TWEEN® 20, for molecular biology, viscous liquid
Roche
Colcemid, solution, suitable for blocking, sterile; 0.2 μm filtered
Sigma-Aldrich
Eg5 Inhibitor III, Dimethylenastron, The Eg5 Inhibitor III, Dimethylenastron, also referenced under CAS 863774-58-7, controls the biological activity of Eg5. This small molecule/inhibitor is primarily used for Membrane applications.
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Monoclonal Anti-β-Tubulin antibody produced in mouse, clone TUB 2.1, ascites fluid
Sigma-Aldrich
Doxycycline hydrochloride
Sigma-Aldrich
MG-132, A cell-permeable, potent, reversible proteasome inhibitor (Ki = 4 nM).