Merck
Cytotoxic flavonoids from the leaves of Cryptocarya chinensis.
Journal of natural products (2010-08-14)
Tsung-Hsien Chou, Jih-Jung Chen, Shiow-Ju Lee, Michael Y Chiang, Cheng-Wei Yang, Ih-Sheng Chen
ABSTRACT
Bioassay-guided fractionation led to the isolation of six new tetrahydroflavanones, cryptochinones A-F (1-6), from the neutral CHCl(3) fraction of Cryptocarya chinensis leaves, together with 14 known compounds (7-20). The structures of these new compounds were determined through spectroscopic analyses, including 2D-NMR, MS, CD, and X-ray crystallographic analysis. Among the isolates, infectocaryone (7) showed cytotoxic activities with IC(50) values of 11.0 and 3.7 μM against NCI-H460 and SF-268 cell lines, respectively, and cryptocaryanone A (9) showed cytotoxic activities with IC(50) values of 5.1, 4.3, and 5.0 μM against MCF-7, NCI-H460, and SF-268 cell lines, respectively.
Product Description
Supelco
Mettler-Toledo Calibration substance ME 51143093, Vanillin, traceable to primary standards (LGC)
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ≥95% (HPLC)
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ~98% (HPLC)
Sigma-Aldrich
Vanillin, tested according to Ph. Eur.
Sigma-Aldrich
Vanillin, ≥97%, FCC, FG
Sigma-Aldrich
Vanillin, natural, ≥97%, FCC, FG
Sigma-Aldrich
Vanillin, ReagentPlus®, 99%
Sigma-Aldrich
Squalene, ≥98%, liquid