Merck
  • Foreign travel is a major risk factor for colonization with Escherichia coli producing CTX-M-type extended-spectrum beta-lactamases: a prospective study with Swedish volunteers.
Foreign travel is a major risk factor for colonization with Escherichia coli producing CTX-M-type extended-spectrum beta-lactamases: a prospective study with Swedish volunteers.
Antimicrobial agents and chemotherapy (2010-06-16)
Thomas Tängdén, Otto Cars, Asa Melhus, Elisabeth Löwdin
ABSTRACT
Foreign travel has been suggested to be a risk factor for the acquisition of extended-spectrum beta-lactamase (ESBL)-producing Enterobacteriaceae. To our knowledge, this has not previously been demonstrated in a prospective study. Healthy volunteers traveling outside Northern Europe were enrolled. Rectal swabs and data on potential travel-associated risk factors were collected before and after traveling. A total of 105 volunteers were enrolled. Four of them did not complete the study, and one participant carried ESBL-producing Escherichia coli before travel. Twenty-four of 100 participants with negative pretravel samples were colonized with ESBL-producing Escherichia coli after the trip. All strains produced CTX-M enzymes, mostly CTX-M-15, and some coproduced TEM or SHV enzymes. Coresistance to several antibiotic subclasses was common. Travel to India was associated with the highest risk for the acquisition of ESBLs (88%; n = 7). Gastroenteritis during the trip was an additional risk factor (P = 0.003). Five of 21 volunteers who completed the follow-up after 6 months had persistent colonization with ESBLs. This is the first prospective study demonstrating that international travel is a major risk factor for colonization with ESBL-producing Enterobacteriaceae. Considering the high acquisition rate of 24%, it is obvious that global efforts are needed to meet the emergence and spread of CTX-M enzymes and other antimicrobial resistances.
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Supelco
Nitrofurantoin, VETRANAL®, analytical standard
Sigma-Aldrich
Nitrofurantoin, 98.0-102.0% (EP, UV)