Merck
  • Biological transformation of chlorophyll-rich spinach (Spinacia oleracea L.) extracts under in vitro gastrointestinal digestion and colonic fermentation.
Biological transformation of chlorophyll-rich spinach (Spinacia oleracea L.) extracts under in vitro gastrointestinal digestion and colonic fermentation.
Food research international (Ottawa, Ont.) (2021-01-30)
Yuanyuan Li, Feng Lu, Xiao Wang, Xiaosong Hu, Xiaojun Liao, Yan Zhang
ABSTRACT
Chlorophyll can be obtained from a variety of green vegetables. In this study, chlorophyll-rich spinach (Spinacia oleracea L.) extracts were subjected to early-life and adult-life gastrointestinal digestion and colonic fermentation in a murine model in vitro to investigate the biological transformation of the chlorophyll. Chlorophylls a and b were the main compounds present in the extracts. Furthermore, some other compounds were also confirmed, such as 151-hydroxy-lactone chlorophyll a, 132-hydroxy chlorophyll a, and 151-hydroxy-lactone chlorophyll b. Chlorophylls favored pheophytinization and oxidative reactions under in vitro early-life and adult-life gastrointestinal digestion, leading to the formation of pheophytin a, pheophytin b, 132-hydroxy pheophytin a, and 151-hydroxy-lactone pheophytin a. 16S rRNA gene sequencing conveyed that pheophytins modulated the gut microbiota composition during in vitro colonic fermentation. Notably, Blautia in the gut microbiota of 3-week-old mice (early life) and unclassified Lachnospiraceae in 8-week-old mice (adult life) were advantageous for transforming the pheophytins to pheophorbide a, pheophorbide b, 151-hydroxy-lactone pheophorbide a, and 132-hydroxy pheophorbide a, thereby demonstrating the loss of the phytol chain in the pheophytins. Meanwhile, total short-chain fatty acids, as well as acetic, propionic, and butyric acids, were increased by the process of microbial fermentation in the presence of pheophytins. Our study provides fundamental insight into the contribution of diverse gut microbiota functions toward the biological transformation of pheophytins.
Sigma-Aldrich
Pepsin from porcine gastric mucosa, lyophilized powder, ≥3,200 units/mg protein
Sigma-Aldrich
Lipoprotein Lipase from bovine milk, ammonium sulfate suspension, ≥2,000 units/mg protein (BCA)
Supelco
Chlorophyll a, analytical standard
Supelco
Chlorophyll b, analytical standard
Sigma-Aldrich
Pancreatin from porcine pancreas, 8 × USP specifications
Sigma-Aldrich
Bile extract porcine